Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03057591_g1
          See other CFL1 GE Assays ›
          Gene Symbol:
          Cfl1
          Gene Name:
          cofilin 1, non-muscle
          Gene Aliases:
          AA959946, Cof, n-cofilin
          Chromosome Location:
          Chr.19: 5490455 - 5494031 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03057591_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cfl1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007687.5 NP_031713.1 3 - 4 534 73
          GenBank mRNA AK013943.1 - 3 - 4 463 73
          AK050651.1 - 465 73
          AK088633.1 - 3 - 4 470 73
          AK088793.1 - 3 - 4 469 73
          AK167795.1 - 3 - 4 465 73
          BC046225.1 - 3 - 4 429 5691650 73
          BC058726.1 - 3 - 4 462 5715707 73
          BC094357.1 - 3 - 4 444 6820355 73
          CX218255.1 - 3 - 4 570 73
          D00472.1 - 3 - 4 483 73

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cfl1

          Panther Classification:

          Molecular Function -

          non-motor actin binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic cytokinesis
          neural crest cell migration
          neural fold formation
          protein phosphorylation
          protein import into nucleus
          movement of cell or subcellular component
          cytoskeleton organization
          actin filament organization
          regulation of cell morphogenesis
          establishment of cell polarity
          cell projection organization
          actin filament depolymerization
          positive regulation of actin filament depolymerization
          response to amino acid
          negative regulation of cell size
          regulation of dendritic spine morphogenesis
          actin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31929042 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31929040 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31929038 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31929037 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31929039 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31929041 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31929034 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31929035 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31929036 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2000548 BC046225.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2000548 D00472.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2000548 AK050651.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2000548 AK088633.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2000548 AK195534.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974788 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P748958 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 404944 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 884952 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 13388 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974788 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P748958 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2000548 BC046225.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2000548 D00472.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2000548 AK050651.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2000548 AK088633.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2000548 AK195534.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P748958 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P477720 AK020471.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31929043 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31929037 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31929035 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31929034 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31929056 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31929049 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31929048 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31929058 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31929040 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31929046 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31929051 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31929059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31929038 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09093596 - -
          Whole Mouse Genome G4122A A_52_P748958 - -
          Whole Mouse Genome G4122A A_52_P477720 AK020471.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127610 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136114 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14493577 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31929038 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31929041 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31929042 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31929034 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31929040 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31929039 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31929036 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31929037 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31929035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928997 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31929043 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31929049 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31929034 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14280193 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31929054 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31929030 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31929066 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31929025 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31929058 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14493581 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31929038 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31929027 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14311972 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31929040 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14234502 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31929028 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14280199 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31929056 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14311968 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31929059 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10460618 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460619 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460617 - -
          Affymetrix HT MG-430 PM Array Plate 1455138_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1448346_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10460616 - -
          Mouse Genome 430 2.0 Array 1455138_PM_x_at - -
          Mouse Genome 430 2.0 Array 1448346_PM_at - -
          Mouse Genome 430 2.0 Array 1448346_at - -
          Mouse Genome 430 2.0 Array 1455138_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12631_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12631_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1900000897.mm.1 NM_007687.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000897.mm.1 NM_007687.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000897.mm.1 TC1900000073.mm.1 MTA TC
          Mouse Expression Set 430 1455138_x_at - -
          Mouse Expression Set 430 1455138_PM_x_at - -
          Mouse Expression Set 430 1448346_PM_at - -
          Mouse Expression Set 430 1448346_at - -
          Murine Genome U74v2 Set 99119_at - -
          Murine Genome U74v2 Set 99119_PM_at - -
          Mouse Genome 430A 2.0 Array 1448346_at - -
          Mouse Genome 430A 2.0 Array 1455138_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1448346_PM_at - -
          Mouse Genome 430A 2.0 Array 1455138_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1455138_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1448346_at - -
          Affymetrix HT Mouse Genome 430A Array 1455138_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1448346_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 523689 - -
          ABI Mouse Genome Survey Microarray 542100 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2340735 - -
          Illumina mouse-6 v1.0 expression beadchip 2340735 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002340735 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1213376 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1213376 - -
          mouse-6 v1.1 (Illumina) scl0012631.1_129-S - -
          Illumina Sentrix mouse-6.v1 bead array 2340735 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0012631.1_129-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2340735 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1213376 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1213376 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0012631.1_129-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline