Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03060489_uH
          See other SSXB3 GE Assays ›
          Gene Symbol:
          Ssxb3 Ssxb11 Ssxb2 Ssxb8 Ssxb9
          Gene Name:
          synovial sarcoma, X member B, breakpoint 3
          synovial sarcoma, X member B, breakpoint 11
          synovial sarcoma, X member B, breakpoint 2
          synovial sarcoma, X member B, breakpoint 8
          synovial sarcoma, X member B, breakpoint 9
          Gene Aliases:
          -
          -
          -
          -
          -
          Chromosome Location:
          Chr.X: 8583404 - 8591810 on Build GRCm38
          Chromosome Location:
          -
          Chromosome Location:
          Chr.X: 8454343 - 8461726 on Build GRCm38
          Chromosome Location:
          Chr.X: 8684843 - 8690883 on Build GRCm38
          Chromosome Location:
          Chr.X: 8366978 - 8375388 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          Assay ID Mm03060489_uH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ssxb3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_198898.2 NP_942601.1 2 162 134
          GenBank mRNA AY244794.1 - 2 162 134
          BC087939.1 - 3 242 134
          BC132618.1 - 2 162 40130895 134
          BC132644.1 - 2 162 40130921 134
          Gene Symbol: Ssxb11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AY244802.1 - 2 161 134
          Gene Symbol: Ssxb2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001001450.4 NP_001001450.1 4 474 134
          NM_001134226.1 NP_001127698.1 3 297 134
          GenBank mRNA AK076879.1 - 4 475 134
          AY244793.1 - 4 475 134
          BC048441.1 - 3 297 6773922 134
          BC089478.1 - 3 297 6743842 134
          Gene Symbol: Ssxb8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001081565.3 NP_001075034.3 2 121 134
          Gene Symbol: Ssxb9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_199066.2 NP_951021.2 3 298 134
          GenBank mRNA AY244800.1 - 298 134

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Amplicon spans exons and probe does not span exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ssxb3
          Click here to find siRNAs for Gene Ssxb11
          Click here to find siRNAs for Gene Ssxb2
          Click here to find siRNAs for Gene Ssxb8
          Click here to find siRNAs for Gene Ssxb9

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0X00011610.mm.1 NM_001001450.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011610.mm.1 NM_001001450.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011610.mm.1 TC0X00001802.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00011611.mm.1 NM_001001450.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011611.mm.1 NM_001001450.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011611.mm.1 NM_001134226.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011611.mm.1 NM_001134226.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011611.mm.1 TC0X00001802.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00011632.mm.1 NM_198898.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011632.mm.1 NM_198898.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011632.mm.1 TC0X00003415.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00011608.mm.1 NM_001001450.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011608.mm.1 NM_001001450.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011608.mm.1 NM_001134226.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011608.mm.1 NM_001134226.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011608.mm.1 TC0X00001802.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00011631.mm.1 NM_198898.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011631.mm.1 NM_198898.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011631.mm.1 TC0X00003415.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00011643.mm.1 NM_001081565.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011643.mm.1 NM_001081565.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011643.mm.1 TC0X00003416.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00011609.mm.1 NM_001001450.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011609.mm.1 NM_001001450.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011609.mm.1 NM_001134226.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011609.mm.1 NM_001134226.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011609.mm.1 TC0X00001802.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00011633.mm.1 NM_198898.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011633.mm.1 NM_198898.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011633.mm.1 TC0X00003415.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00011641.mm.1 NM_001081565.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011641.mm.1 NM_001081565.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011641.mm.1 TC0X00003416.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00011642.mm.1 NM_001081565.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011642.mm.1 NM_001081565.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011642.mm.1 TC0X00003416.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00000694.mm.1 NM_199066.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00000694.mm.1 NM_199066.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00000694.mm.1 TC0X00003384.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 426996 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-9kpsn:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0