Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03061419_gH
          See other DCPP2 GE Assays ›
          Gene Symbol:
          Dcpp2
          Gene Name:
          demilune cell and parotid protein 2
          Gene Aliases:
          Dcpp-2, EG630537, SPT-2
          Chromosome Location:
          Chr.17: 23898722 - 23900787 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03061419_gH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dcpp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001039238.2 NP_001034327.2 4 324 128

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2158510 NM_001039238.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158510 NM_001039238.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094265 - -
          Whole Mouse Genome G4122A A_52_P535962 BC059096.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P535962 BC171996.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13244080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094297 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13550422 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094302 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13154595 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13185898 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13445449 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13081961 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094309 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094306 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13056984 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094297 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094265 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10442374 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442372 - -
          Affymetrix Mouse Gene 1.1 ST Array 10442370 - -
          Affymetrix Mouse Gene 1.1 ST Array 10442373 - -
          Affymetrix Mouse Clariom S Array PSR1700002406.mm.1 NM_001039238.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002406.mm.1 NM_001039238.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002406.mm.1 TC1700000327.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700002408.mm.1 NM_001039238.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002408.mm.1 NM_001039238.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002408.mm.1 TC1700000327.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700002407.mm.1 NM_001039238.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002407.mm.1 NM_001039238.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002407.mm.1 TC1700000327.mm.1 MTA TC
          Mouse Expression Set 430 1459725_s_at - -
          Mouse Expression Set 430 1459725_PM_s_at - -
          Murine Genome U74v2 Set 93765_PM_at - -
          Murine Genome U74v2 Set 93765_at - -
          Mouse Genome 430A 2.0 Array 1459725_s_at - -
          Mouse Genome 430A 2.0 Array 1459725_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1221839 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2816271 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2816271 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline