Search
Search

Klra7| Gene Symbol: Klra7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110323.1 | NP_001103793.1 | 3 - 4 | 331 | 151 | |
| NM_014194.5 | NP_055009.5 | 2 - 3 | 271 | 151 | ||
| GenBank mRNA | AF074457.1 | - | 2 - 3 | 208 | 151 | |
| AF283248.1 | - | 2 - 3 | 208 | 151 | ||
| AF283249.1 | - | 2 - 3 | 208 | 151 | ||
| AF288375.1 | - | 2 - 3 | 201 | 151 | ||
| AF288376.1 | - | 2 - 3 | 230 | 151 | ||
| AF307945.1 | - | 2 - 3 | 208 | 151 | ||
| AF307946.1 | - | 2 - 3 | 208 | 151 | ||
| AF419249.1 | - | 490 | 151 | |||
| AK143240.1 | - | 2 - 3 | 231 | 150 | ||
| BC099859.1 | - | 2 - 3 | 207 | 40040027 | 151 | |
| BC099860.1 | - | 2 - 3 | 207 | 40040024 | 151 | |
| BC099862.1 | - | 2 - 3 | 207 | 40040021 | 151 | |
| BC130251.1 | - | 2 - 3 | 256 | 40142464 | 151 | |
| BC140984.1 | - | 2 - 3 | 254 | 9055596 | 151 | |
| EU429321.1 | - | 2 - 3 | 201 | 151 | ||
| GU434662.1 | - | 2 - 3 | 201 | 151 | ||
| GU434663.1 | - | 2 - 3 | 201 | 151 | ||
| S78689.1 | - | 2 - 3 | 230 | 151 | ||
| U10093.1 | - | 3 - 4 | 331 | 151 | ||
| U10094.1 | - | 2 - 3 | 271 | 151 | ||
| U10095.1 | - | 3 - 4 | 331 | 151 | ||
| U12890.1 | - | 2 - 3 | 222 | 151 | ||
Set Membership: |
|