Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03954196_s1
          See other 2900064F13RIK GE Assays ›
          Gene Symbol:
          2900064F13Rik
          Gene Name:
          RIKEN cDNA 2900064F13 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03954196_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2900064F13Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK013749.1 - 1 247 78

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream phosphoprotein enriched in astrocytes 15B Pea15b -4.3mb Anti-sense Chr. 5 - 77510039 - 77511003 on NCBI Build GRCm38
          Downstream RIKEN cDNA 1700031L13 gene 1700031L13Rik 294.9kb Anti-sense Chr. 5 - 82122693 - 82124711 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2415930 AK013749.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2415930 AV154628.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 427448 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 54866 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2415930 AK013749.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2415930 AV154628.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1034835 AK013749.1 mouse_transcriptome
          Agilent Six_ChIP_01_of_03_v2 A_68_P23842787 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04339166 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23842802 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23842818 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23842816 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23842808 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23842800 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23842805 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23842819 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23842827 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23842825 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23842804 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04420509 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23842774 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04420507 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23842817 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04284985 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23842788 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04492508 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23842771 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23842786 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23842776 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23842821 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23842824 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23842793 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23842813 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23842790 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04339162 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23842811 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23842807 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23842822 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23842812 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23842794 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04492509 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23842782 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1432813_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1432814_PM_at - -
          Mouse Genome 430 2.0 Array 1432814_at - -
          Mouse Genome 430 2.0 Array 1432813_at - -
          Mouse Genome 430 2.0 Array 1432813_PM_at - -
          Mouse Genome 430 2.0 Array 1432814_PM_at - -
          Mouse Expression Set 430 1432813_at - -
          Mouse Expression Set 430 1432814_PM_at - -
          Mouse Expression Set 430 1432814_at - -
          Mouse Expression Set 430 1432813_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432813_at - -
          Affymetrix HT Mouse Genome 430B Array 1432813_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432814_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432814_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 436497 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-bkzv6:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0