Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04205882_gH
          See other OLFR192 GE Assays ›
          Gene Symbol:
          Olfr192 Olfr190
          Gene Name:
          olfactory receptor 192
          olfactory receptor 190
          Gene Aliases:
          -
          MOR183-4
          Chromosome Location:
          Chr.16: 59098066 - 59098990 on Build GRCm38
          Chromosome Location:
          Chr.16: 59074155 - 59075078 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Mm04205882_gH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr192
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_207549.1 NP_997432.1 1 846 102
          Gene Symbol: Olfr190
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BC104050.1 - 1 40040225 102
          BC104051.1 - 1 40040226 102

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr192
          Click here to find siRNAs for Gene Olfr190

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794335 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794325 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794355 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794354 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12871797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794354 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794365 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794355 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794345 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794354 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794334 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794325 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12597563 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794372 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794335 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10440161 - -
          Affymetrix Mouse Gene 1.1 ST Array 10440160 - -
          Affymetrix Mouse Clariom S Array PSR1600014460.mm.1 NM_207549.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600014460.mm.1 NM_207549.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600014460.mm.1 TC1600001790.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 595424 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4210435 - -
          Illumina mouse-6 v1.0 expression beadchip 4210435 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004210435 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2609698 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2609698 - -
          mouse-6 v1.1 (Illumina) scl0404309.1_74-S - -
          Illumina Sentrix mouse-6.v1 bead array 4210435 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0404309.1_74-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4210435 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2609698 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2609698 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-5xwv5:80/100.66.76.242:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0