Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04212296_gH
          See other ZFP747 GE Assays ›
          Gene Symbol:
          Zfp747
          Gene Name:
          zinc finger protein 747
          Gene Aliases:
          6430604K15Rik
          Chromosome Location:
          Chr.7: 127372537 - 127376050 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04212296_gH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp747
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175560.3 NP_780769.1 2 - 3 522 112
          GenBank mRNA AK032584.1 - 2 - 3 488 112
          AK042972.1 - 2 - 3 523 112
          BC058799.1 - 2 - 3 468 6484930 112

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp747

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25505328 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25505329 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25505387 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25505327 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25505330 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2002465 AK032584.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2002465 AK042972.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2002465 NM_175560.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2002466 AK032584.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2002466 AK042972.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2002466 NM_175560.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2002466 AK032584.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2002466 AK042972.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2002466 NM_175560.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2002465 AK032584.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2002465 AK042972.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2002465 NM_175560.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505387 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06361494 - -
          Whole Mouse Genome G4122A A_51_P242608 AK088474.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P242608 BC069858.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P242608 NM_198011.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140642 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135583 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06517285 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25505327 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25505387 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25505330 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25505328 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25505329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505339 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06517276 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505353 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505350 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06381335 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505361 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505369 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505348 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505355 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10568230 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568232 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568231 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568229 - -
          Affymetrix Mouse Gene 1.1 ST Array 10568228 - -
          Affymetrix Mouse Clariom S Array PSR0700036304.mm.1 NM_175560.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036304.mm.1 NM_175560.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036304.mm.1 TC0700004324.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700036305.mm.1 NM_175560.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036305.mm.1 NM_175560.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036305.mm.1 TC0700004324.mm.1 MTA TC
          Mouse Expression Set 430 1451571_s_at - -
          Mouse Expression Set 430 1451571_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1451571_s_at - -
          Mouse Genome 430A 2.0 Array 1451571_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 629116 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5290041 - -
          Illumina mouse-6 v1.0 expression beadchip 5290041 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005290041 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1219375 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1219375 - -
          mouse-6 v1.1 (Illumina) scl0269997.1_231-S - -
          Illumina Sentrix mouse-6.v1 bead array 5290041 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0269997.1_231-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5290041 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1219375 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1219375 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline