Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04214367_g1
          See other GM1140 GE Assays ›
          Gene Symbol:
          Gm1140
          Gene Name:
          predicted gene 1140
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 67682265 - 67694458 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04214367_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm1140
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001126317.1 NP_001119789.1 8 654 100

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Amplicon spans exons and probe does not span exons

          Gene Ontology Categories:

          Function(s) Process(es)

          spermatid development
          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2107978 NM_001126317.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2107978 NM_001126317.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32437565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32437560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32437651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32437563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32437740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32437557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32437562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32437559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32437738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32437736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32437566 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32437651 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15132882 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1446051_PM_at - -
          Mouse Genome 430 2.0 Array 1446051_at - -
          Mouse Genome 430 2.0 Array 1446051_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0X00003816.mm.1 NM_001126317.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003816.mm.1 NM_001163195.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003816.mm.1 NM_001163195.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003816.mm.1 TC0X00000605.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00003817.mm.1 NM_001126317.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003817.mm.1 TC0X00000605.mm.1 MTA TC
          Mouse Expression Set 430 1446051_PM_at - -
          Mouse Expression Set 430 1446051_at - -
          Affymetrix HT Mouse Genome 430B Array 1446051_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1446051_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 685079 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline