Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04337260_g1
          See other E030030I06RIK GE Assays ›
          Gene Symbol:
          E030030I06Rik
          Gene Name:
          RIKEN cDNA E030030I06 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 22113050 - 22149270 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04337260_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: E030030I06Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001254744.1 NP_001241673.1 2 1909 102
          GenBank mRNA AK045260.1 - 2 1459 102

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2150283 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2150299 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P547764 AK045260.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P547764 AK045260.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P109320 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P101020 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2150283 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2150299 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P547764 AK045260.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P547764 AK045260.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P109320 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02082468 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02082470 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P101020 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06999511 - -
          Whole Mouse Genome G4122A A_52_P547764 AK045260.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P547764 AK045260.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120091 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P02082470 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P109320 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P101020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26997073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26997070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26997052 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26997042 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1442156_PM_at - -
          Mouse Genome 430 2.0 Array 1442156_at - -
          Mouse Genome 430 2.0 Array 1442156_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 319887_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 319887_at - -
          Affymetrix Mouse Clariom S Array PSR1000013409.mm.1 NM_001254744.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013409.mm.1 NM_001254744.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013409.mm.1 NM_001254745.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013409.mm.1 NM_001254745.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013409.mm.1 TC1000001851.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000013413.mm.1 NM_001254744.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013413.mm.1 NM_001254744.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013413.mm.1 NM_001254745.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013413.mm.1 NM_001254745.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013413.mm.1 TC1000001851.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000013402.mm.1 NM_001254744.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013402.mm.1 NM_001254744.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013402.mm.1 TC1000001851.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000013408.mm.1 NM_001254744.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013408.mm.1 NM_001254744.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013408.mm.1 NM_001254745.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013408.mm.1 NM_001254745.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013408.mm.1 TC1000001851.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000013410.mm.1 NM_001254744.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013410.mm.1 NM_001254744.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013410.mm.1 NM_001254745.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013410.mm.1 NM_001254745.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013410.mm.1 TC1000001851.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000013412.mm.1 NM_001254744.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013412.mm.1 NM_001254744.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013412.mm.1 NM_001254745.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013412.mm.1 NM_001254745.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013412.mm.1 TC1000001851.mm.1 MTA TC
          Mouse Expression Set 430 1442156_PM_at - -
          Mouse Expression Set 430 1442156_at - -
          Affymetrix HT Mouse Genome 430B Array 1442156_at - -
          Affymetrix HT Mouse Genome 430B Array 1442156_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 342988 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6130112 - -
          MouseWG-6 v2 Expression BeadChip 3780605 - -
          MouseWG-6 v2 Expression BeadChip 430082 - -
          Illumina mouse-6 v1.0 expression beadchip 430082 - -
          Illumina mouse-6 v1.0 expression beadchip 3780605 - -
          Illumina mouse-6 v1.0 expression beadchip 6130112 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006130112 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003780605 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000430082 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1236476 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1248267 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1259231 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1259231 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1236476 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1248267 - -
          mouse-6 v1.1 (Illumina) scl0319887.1_156-S - -
          mouse-6 v1.1 (Illumina) scl00319887.1_120-S - -
          mouse-6 v1.1 (Illumina) scl0319887.1_22-S - -
          Illumina Sentrix mouse-6.v1 bead array 430082 - -
          Illumina Sentrix mouse-6.v1 bead array 6130112 - -
          Illumina Sentrix mouse-6.v1 bead array 3780605 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0319887.1_156-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0319887.1_22-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00319887.1_120-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6130112 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 430082 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3780605 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1248267 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1259231 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2650420 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline