Search
Search

CXCL11| Gene Symbol: CXCL11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032950.1 | NP_001028122.1 | 190 | 106 | ||
| XM_028847939.1 | XP_028703772.1 | 424 | 106 | |||
| GenBank mRNA | AY044447.1 | - | 190 | 106 | ||
Set Membership: |
|