Search
Search

KMT2A| Gene Symbol: KMT2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015115825.2 | XP_014971311.2 | 11091 | 84 | ||
| XM_015115826.2 | XP_014971312.2 | 11082 | 84 | |||
| XM_028834035.1 | XP_028689868.1 | 11179 | 84 | |||
| XM_028834036.1 | XP_028689869.1 | 8575 | 84 | |||
| GenBank mRNA | JU343756.1 | - | 10646 | 84 | ||
| JU486874.1 | - | 10658 | 84 | |||
Set Membership: |
|