Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00440945_m1
          See other PPARG GE Assays ›
          Gene Symbol:
          Pparg
          Gene Name:
          peroxisome proliferator-activated receptor gamma
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 147274057 - 147399383 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00440945_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (2)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pparg
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001145366.1 NP_001138838.1 7 - 8 1383 105
          NM_001145367.1 NP_001138839.1 7 - 8 1561 105
          NM_013124.3 NP_037256.1 6 - 7 1358 105
          XM_006237009.2 XP_006237071.1 7 - 8 1354 105
          GenBank mRNA AB011365.1 - 5 - 6 1191 105
          AB019561.1 - 6 - 7 1272 105
          AF156665.1 - 7 - 8 1383 105
          AF156666.1 - 6 - 7 1320 105
          AF246457.1 - 7 - 8 1311 105
          AF246458.1 - 7 - 8 1351 105
          Y12882.2 - 6 - 7 1321 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400001603.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Nuclear Receptor 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pparg

          Panther Classification:

          Molecular Function -

          C4 zinc finger nuclear receptor DNA-binding transcription factor zinc finger transcription factor gene-specific transcriptional regulator

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          negative regulation of cytokine production
          placenta development
          negative regulation of acute inflammatory response
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          fatty acid metabolic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          signal transduction
          heart development
          regulation of blood pressure
          negative regulation of cell proliferation
          response to cold
          response to light stimulus
          response to mechanical stimulus
          response to organic substance
          negative regulation of macrophage derived foam cell differentiation
          negative regulation of receptor biosynthetic process
          negative regulation of cholesterol storage
          negative regulation of sequestering of triglyceride
          response to organic cyclic compound
          long-chain fatty acid transport
          regulation of lipid metabolic process
          fatty acid oxidation
          monocyte differentiation
          negative regulation of cell growth
          epithelial cell differentiation
          response to caffeine
          organ regeneration
          response to food
          response to retinoic acid
          cellular response to insulin stimulus
          negative regulation of collagen biosynthetic process
          response to vitamin A
          response to lipid
          peroxisome proliferator activated receptor signaling pathway
          response to immobilization stress
          response to diuretic
          response to drug
          glucose homeostasis
          response to starvation
          regulation of circadian rhythm
          lipoprotein transport
          positive regulation of apoptotic process
          steroid hormone mediated signaling pathway
          response to estrogen
          cell fate commitment
          fat cell differentiation
          regulation of fat cell differentiation
          positive regulation of fat cell differentiation
          low-density lipoprotein particle receptor biosynthetic process
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of fatty acid oxidation
          cell maturation
          rhythmic process
          negative regulation of smooth muscle cell proliferation
          positive regulation of oligodendrocyte differentiation
          white fat cell differentiation
          brown fat cell differentiation
          positive regulation of sequence-specific DNA binding transcription factor activity
          negative regulation of telomerase activity
          response to low-density lipoprotein particle
          positive regulation of phagocytosis, engulfment
          negative regulation of interferon-gamma-mediated signaling pathway
          regulation of transcription involved in cell fate commitment
          cellular response to lithium ion
          cellular response to retinoic acid
          cellular response to vitamin E
          cellular response to prostaglandin stimulus
          cellular response to prostaglandin E stimulus
          cellular response to organic cyclic compound
          cellular response to hyperoxia
          negative regulation of peptide hormone secretion
          negative regulation of cellular response to insulin stimulus
          response to metformin
          negative regulation of pancreatic stellate cell proliferation
          RNA polymerase II regulatory region DNA binding
          core promoter sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          DNA binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          steroid hormone receptor activity
          RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
          protein binding
          transcription factor binding
          drug binding
          zinc ion binding
          enzyme binding
          protein phosphatase binding
          estrogen receptor binding
          ligand-dependent nuclear receptor transcription coactivator activity
          activating transcription factor binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          retinoid X receptor binding
          arachidonic acid binding
          alpha-actinin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P229432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329486 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472095 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90916 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772379 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 151825 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16188 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42532 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 169510 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78611 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56181 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58929 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108315 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 155858 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 173026 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142610 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62239 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113548 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P229432 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P229432 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P229432 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P123333 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700628022 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79093 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39467 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4566 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38052 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39332 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63093 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41029 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97745 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47112 - -
          Agilent Rat cDNA Microarray (G4105A) 700628022 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10857991 - -
          Affymetrix Rat Gene 1.0 ST Array 10857998 - -
          Affymetrix Rat Gene 1.0 ST Array 10857995 - -
          Affymetrix Rat Gene 1.0 ST Array 10857986 - -
          Affymetrix Rat Gene 1.0 ST Array 10857994 - -
          Affymetrix Rat Gene 1.0 ST Array 10857997 - -
          Affymetrix Rat Gene 1.0 ST Array 10857993 - -
          Affymetrix Rat Gene 1.0 ST Array 10857996 - -
          Affymetrix Rat Gene 1.0 ST Array 10857992 - -
          Affymetrix Rat Gene 1.0 ST Array 10857987 - -
          Affymetrix Rat Gene 1.0 ST Array 10857988 - -
          Affymetrix Rat Gene 1.0 ST Array 10857985 - -
          Affymetrix Rat Gene 1.0 ST Array 10857990 - -
          Affymetrix Rat Gene 1.0 ST Array 10857989 - -
          Affymetrix Rat Gene 1.1 ST Array 10857984 - -
          Rat Genome 230 2.0 Array 1369179_a_at - -
          Rat Genome 230 2.0 Array 1369179_PM_a_at - -
          Affymetrix HT Rat Focus Array 1369179_a_at - -
          Affymetrix HT Rat Focus Array 1369179_PM_a_at - -
          Affymetrix Rat Clariom S Assay PSR0400011435.rn.1 TC0400001603.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400011436.rn.1 TC0400001603.rn.1 RTA TC
          Rat Expression Set 230 1369179_a_at - -
          Rat Expression Set 230 1369179_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10857984 - -
          Rat Genome U34 Set AB011365_s_at - -
          Rat Genome U34 Set rc_AI012992_s_at - -
          Rat Genome U34 Set AB011365_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21773923 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372572 - -
          RatRef-12 Expression BeadChip ILMN_48832 - -

          Back To Top
           

          Citations (2)

          Causes and prevention of tamoxifen-induced accumulation of triacylglycerol in rat liver.
          Authors
          Gudbrandsen OA, Rost TH, Berge RK
          Journal
          J Lipid Res - Page No 2223 (Sun Oct 01 00:00:00 EDT 2006)
          Volume (Year)
          47 (2006)
          Product Usage
          -
          Early postnatal oestradiol exposure causes insulin resistance and signs of inflammation in circulation and skeletal muscle.
          Authors
          Alexanderson C, Eriksson E, Stener-Victorin E, Lonn M, Holmang A
          Journal
          J Endocrinol - Page No 49 (Wed Apr 01 00:00:00 EDT 2009)
          Volume (Year)
          201 (2009)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline