Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00442194_m1
          See other AKT3 GE Assays ›
          Gene Symbol:
          Akt3
          Gene Name:
          v-akt murine thymoma viral oncogene homolog 3
          Gene Aliases:
          Pkbg
          Chromosome Location:
          Chr.13: 95076308 - 95348913 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00442194_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Akt3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031575.1 NP_113763.1 2 - 3 217 71
          GenBank mRNA D49836.1 - 2 - 3 217 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300002011.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Akt3

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          mitochondrial genome maintenance
          protein phosphorylation
          signal transduction
          positive regulation of sodium ion transport
          cellular response to insulin stimulus
          protein serine/threonine kinase activity
          protein kinase C binding
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P286853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 434960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 743838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922648 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700360 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131140 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 322853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 765827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345095 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 420006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673674 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 696450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416414 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942179 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 81457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750894 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203050 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 675629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 322861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 578389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781389 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3495 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136624 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138978 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85915 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121478 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62922 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126137 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8111 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134399 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100036 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 11668 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13957 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69582 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 28174 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145921 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175498 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134987 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 155748 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24093 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82155 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146211 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87141 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143478 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43209 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 123571 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114549 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 160340 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P286853 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P286853 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P286853 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P286853 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700811104 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70120 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82816 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50170 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87010 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56077 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18976 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 101007 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85980 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103612 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11870 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60198 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30716 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35225 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82464 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73120 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65540 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56954 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98763 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66057 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44487 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70299 - -
          Agilent Rat cDNA Microarray (G4105A) 700811104 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10770206 - -
          Affymetrix Rat Gene 1.0 ST Array 10770200 - -
          Affymetrix Rat Gene 1.0 ST Array 10770201 - -
          Affymetrix Rat Gene 1.0 ST Array 10770205 - -
          Affymetrix Rat Gene 1.0 ST Array 10770204 - -
          Affymetrix Rat Gene 1.0 ST Array 10770207 - -
          Affymetrix Rat Gene 1.0 ST Array 10770203 - -
          Affymetrix Rat Gene 1.0 ST Array 10770202 - -
          Affymetrix Rat Gene 1.1 ST Array 10770197 - -
          Rat Genome 230 2.0 Array 1387592_at - -
          Rat Genome 230 2.0 Array 1387592_PM_at - -
          Rat Genome 230 2.0 Array 1372874_at - -
          Rat Genome 230 2.0 Array 1372874_PM_at - -
          Affymetrix HT Rat Focus Array 1387592_PM_at - -
          Affymetrix HT Rat Focus Array 1387592_at - -
          Affymetrix Rat Clariom S Assay PSR1300014384.rn.1 TC1300002011.rn.1 RTA TC
          Rat Expression Set 230 1387592_PM_at - -
          Rat Expression Set 230 1372874_at - -
          Rat Expression Set 230 1372874_PM_at - -
          Rat Expression Set 230 1387592_at - -
          Affymetrix Rat Gene 1.0 ST Array 10770197 - -
          Rat Genome U34 Set D49836_at - -
          Rat Genome U34 Set rc_AI073194_at - -
          Rat Genome U34 Set D49836_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20826580 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370315 - -
          RatRef-12 Expression BeadChip ILMN_60767 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline