Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00461076_m1
          See other PEX6 GE Assays ›
          Gene Symbol:
          Pex6
          Gene Name:
          peroxisomal biogenesis factor 6
          Gene Aliases:
          Paf-2, Paf2
          Chromosome Location:
          Chr.9: 16568743 - 16580900 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00461076_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pex6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_057125.1 NP_476466.1 3 - 4 1152 70
          GenBank mRNA D63673.1 - 3 - 4 1152 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900001560.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pex6

          Gene Ontology Categories:

          Function(s) Process(es)

          protein targeting to peroxisome
          peroxisome organization
          metabolic process
          protein import into peroxisome matrix, translocation
          protein stabilization
          ATP binding
          protein C-terminus binding
          ATPase activity
          protein complex binding
          ATPase activity, coupled

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P191941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224308 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514013 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120896 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P191941 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P191941 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P191941 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P191941 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2651 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10921679 - -
          Affymetrix Rat Gene 1.0 ST Array 10926480 - -
          Affymetrix Rat Gene 1.0 ST Array 10926473 - -
          Affymetrix Rat Gene 1.0 ST Array 10926472 - -
          Affymetrix Rat Gene 1.0 ST Array 10926478 - -
          Affymetrix Rat Gene 1.0 ST Array 10926477 - -
          Affymetrix Rat Gene 1.0 ST Array 10926481 - -
          Affymetrix Rat Gene 1.0 ST Array 10926479 - -
          Affymetrix Rat Gene 1.0 ST Array 10926484 - -
          Affymetrix Rat Gene 1.0 ST Array 10926475 - -
          Affymetrix Rat Gene 1.0 ST Array 10926474 - -
          Affymetrix Rat Gene 1.0 ST Array 10926471 - -
          Affymetrix Rat Gene 1.0 ST Array 10921678 - -
          Affymetrix Rat Gene 1.0 ST Array 10926476 - -
          Affymetrix Rat Gene 1.0 ST Array 10926482 - -
          Affymetrix Rat Gene 1.0 ST Array 10926483 - -
          Affymetrix Rat Gene 1.1 ST Array 10921677 - -
          Affymetrix Rat Gene 1.1 ST Array 10926470 - -
          Rat Genome 230 2.0 Array 1368264_at - -
          Rat Genome 230 2.0 Array 1368264_PM_at - -
          Affymetrix HT Rat Focus Array 1368264_at - -
          Affymetrix HT Rat Focus Array 1368264_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0900012971.rn.1 TC0900001560.rn.1 RTA TC
          Rat Expression Set 230 1368264_at - -
          Rat Expression Set 230 1368264_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10926470 - -
          Affymetrix Rat Gene 1.0 ST Array 10921677 - -
          Rat Genome U34 Set D63673_at - -
          Rat Genome U34 Set D63673_g_at - -
          Rat Genome U34 Set rc_AI137451_s_at - -
          Rat Genome U34 Set D63673_at - -
          Rat Genome U34 Set D63673_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20914841 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376911 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline