Search
Search
| Gene Symbol: Sra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183329.3 | NP_899158.3 | 2 - 3 | 209 | 90 | |
| GenBank mRNA | AY026354.1 | - | 2 - 3 | 157 | 90 | |
| AY542868.1 | - | 2 - 3 | 209 | 90 | ||
| BC091105.1 | - | 2 - 3 | 397 | 7381969 | 90 | |
Set Membership: |
|