Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00493049_m1
          See other L1CAM GE Assays ›
          Gene Symbol:
          L1cam
          Gene Name:
          L1 cell adhesion molecule
          Gene Aliases:
          Hsas, Hyd, NCAML1
          Chromosome Location:
          Chr.X: 156911336 - 156928056 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00493049_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: L1cam
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017345.1 NP_059041.1 6 - 7 790 74
          XM_008773635.1 XP_008771857.1 6 - 7 799 74
          XM_008773636.1 XP_008771858.1 5 - 6 784 74
          XM_008773637.1 XP_008771859.1 5 - 6 784 74
          GenBank mRNA X59149.1 - 6 - 7 790 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00001309.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene L1cam

          Gene Ontology Categories:

          Function(s) Process(es)

          cell adhesion
          homophilic cell adhesion via plasma membrane adhesion molecules
          heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
          leukocyte cell-cell adhesion
          cell surface receptor signaling pathway
          nervous system development
          axon guidance
          positive regulation of cell-cell adhesion
          cell differentiation
          neuron projection development
          cell-cell adhesion mediated by integrin
          homotypic cell-cell adhesion
          positive regulation of calcium-mediated signaling
          cellular response to ethanol
          cellular response to transforming growth factor beta stimulus
          integrin binding
          PDZ domain binding
          sialic acid binding
          identical protein binding
          protein self-association

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P127362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606685 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22913 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P127362 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P127362 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P127362 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P837568 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76512 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10940214 - -
          Affymetrix Rat Gene 1.0 ST Array 10940209 - -
          Affymetrix Rat Gene 1.0 ST Array 10940216 - -
          Affymetrix Rat Gene 1.0 ST Array 10940205 - -
          Affymetrix Rat Gene 1.0 ST Array 10940220 - -
          Affymetrix Rat Gene 1.0 ST Array 10940221 - -
          Affymetrix Rat Gene 1.0 ST Array 10940213 - -
          Affymetrix Rat Gene 1.0 ST Array 10940197 - -
          Affymetrix Rat Gene 1.0 ST Array 10940200 - -
          Affymetrix Rat Gene 1.0 ST Array 10940201 - -
          Affymetrix Rat Gene 1.0 ST Array 10940198 - -
          Affymetrix Rat Gene 1.0 ST Array 10940222 - -
          Affymetrix Rat Gene 1.0 ST Array 10940223 - -
          Affymetrix Rat Gene 1.0 ST Array 10940208 - -
          Affymetrix Rat Gene 1.0 ST Array 10940202 - -
          Affymetrix Rat Gene 1.0 ST Array 10940218 - -
          Affymetrix Rat Gene 1.0 ST Array 10940206 - -
          Affymetrix Rat Gene 1.0 ST Array 10940199 - -
          Affymetrix Rat Gene 1.0 ST Array 10940212 - -
          Affymetrix Rat Gene 1.0 ST Array 10940196 - -
          Affymetrix Rat Gene 1.0 ST Array 10940203 - -
          Affymetrix Rat Gene 1.0 ST Array 10940207 - -
          Affymetrix Rat Gene 1.0 ST Array 10940219 - -
          Affymetrix Rat Gene 1.0 ST Array 10940211 - -
          Affymetrix Rat Gene 1.0 ST Array 10940215 - -
          Affymetrix Rat Gene 1.0 ST Array 10940217 - -
          Affymetrix Rat Gene 1.0 ST Array 10940204 - -
          Affymetrix Rat Gene 1.1 ST Array 10940195 - -
          Rat Genome 230 2.0 Array 1369213_at - -
          Rat Genome 230 2.0 Array 1369213_PM_at - -
          Affymetrix HT Rat Focus Array 1369213_at - -
          Affymetrix HT Rat Focus Array 1369213_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0X00008003.rn.1 TC0X00001309.rn.1 RTA TC
          Rat Expression Set 230 1369213_at - -
          Rat Expression Set 230 1369213_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10940195 - -
          Rat Genome U34 Set X59149_at - -
          Rat Genome U34 Set X59149_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21537699 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376861 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline