Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00516381_m1
          See other GNB3 GE Assays ›
          Gene Symbol:
          Gnb3
          Gene Name:
          guanine nucleotide binding protein (G protein), beta polypeptide 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 157352558 - 157359242 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00516381_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gnb3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021858.3 NP_068630.1 3 - 4 208 79
          XM_006237383.1 XP_006237445.1 3 - 4 1190 79
          GenBank mRNA BC086422.1 - 3 - 4 211 7193131 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400003740.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gnb3

          Panther Classification:

          Molecular Function -

          heterotrimeric G-protein

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          biological_process
          molecular_function
          signal transducer activity
          spectrin binding
          GTPase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P020775 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220263 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172686 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12137 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12137 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12137 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12137 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65295 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10865556 - -
          Affymetrix Rat Gene 1.0 ST Array 10865555 - -
          Affymetrix Rat Gene 1.0 ST Array 10865561 - -
          Affymetrix Rat Gene 1.0 ST Array 10865554 - -
          Affymetrix Rat Gene 1.0 ST Array 10865559 - -
          Affymetrix Rat Gene 1.0 ST Array 10865558 - -
          Affymetrix Rat Gene 1.0 ST Array 10865560 - -
          Affymetrix Rat Gene 1.0 ST Array 10865553 - -
          Affymetrix Rat Gene 1.0 ST Array 10865557 - -
          Affymetrix Rat Gene 1.1 ST Array 10865552 - -
          Rat Genome 230 2.0 Array 1369876_PM_at - -
          Rat Genome 230 2.0 Array 1369876_at - -
          Rat Genome 230 2.0 Array 1395473_PM_at - -
          Rat Genome 230 2.0 Array 1395473_at - -
          Affymetrix HT Rat Focus Array 1395473_PM_at - -
          Affymetrix HT Rat Focus Array 1395473_at - -
          Affymetrix HT Rat Focus Array 1369876_at - -
          Affymetrix HT Rat Focus Array 1369876_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400026531.rn.1 TC0400003740.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400026521.rn.1 TC0400003740.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400026522.rn.1 TC0400003740.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400026528.rn.1 TC0400003740.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400026523.rn.1 TC0400003740.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400026519.rn.1 TC0400003740.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400026524.rn.1 TC0400003740.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400026520.rn.1 TC0400003740.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400026530.rn.1 TC0400003740.rn.1 RTA TC
          Rat Expression Set 230 1369876_at - -
          Rat Expression Set 230 1369876_PM_at - -
          Rat Expression Set 230 1395473_at - -
          Rat Expression Set 230 1395473_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10865552 - -
          Rat Genome U34 Set L29090cds_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20891526 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353416 - -
          RatRef-12 Expression BeadChip ILMN_53966 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline