Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00555260_m1
          See other ZBTB44 GE Assays ›
          Gene Symbol:
          Zbtb44
          Gene Name:
          zinc finger and BTB domain containing 44
          Gene Aliases:
          Btbd15, RGD1308984
          Chromosome Location:
          Chr.8: 32165623 - 32223341 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00555260_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zbtb44
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001034942.1 NP_001030114.1 4 - 5 1427 106
          XM_006242768.2 XP_006242830.1 4 - 5 1614 106
          XM_006242769.2 XP_006242831.1 4 - 5 1560 106
          GenBank mRNA BC104680.1 - 4 - 5 1427 7456912 106

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800000314.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zbtb44

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          DNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P766678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119096 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139334 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72811 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77363 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142612 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 179453 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153344 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P766678 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P766678 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P766678 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P20674 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81820 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66182 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3889 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99885 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10908789 - -
          Affymetrix Rat Gene 1.0 ST Array 10908797 - -
          Affymetrix Rat Gene 1.0 ST Array 10908792 - -
          Affymetrix Rat Gene 1.0 ST Array 10908793 - -
          Affymetrix Rat Gene 1.0 ST Array 10908796 - -
          Affymetrix Rat Gene 1.0 ST Array 10908791 - -
          Affymetrix Rat Gene 1.0 ST Array 10908790 - -
          Affymetrix Rat Gene 1.1 ST Array 10908788 - -
          Rat Genome 230 2.0 Array 1379439_PM_at - -
          Rat Genome 230 2.0 Array 1379439_at - -
          Affymetrix HT Rat Focus Array 1379439_at - -
          Affymetrix HT Rat Focus Array 1379439_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0800002107.rn.1 TC0800000314.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800002109.rn.1 TC0800000314.rn.1 RTA TC
          Rat Expression Set 230 1379439_at - -
          Rat Expression Set 230 1379439_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10908788 - -
          Rat Genome U34 Set rc_AA850609_at - -
          Rat Genome U34 Set rc_AA925920_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21320666 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365047 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline