Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00560632_m1
          See other ADD2 GE Assays ›
          Gene Symbol:
          Add2
          Gene Name:
          adducin 2 (beta)
          Gene Aliases:
          beta-ADD
          Chromosome Location:
          Chr.4: 117744075 - 117882427 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00560632_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Add2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109880.1 NP_001103350.1 8 - 9 1035 92
          NM_012491.2 NP_036623.1 8 - 9 1035 92
          GenBank mRNA M63894.1 - 8 - 9 1035 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400001305.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Add2

          Panther Classification:

          Molecular Function -

          non-motor actin binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          protein complex assembly
          ion transport
          hemopoiesis
          positive regulation of protein binding
          barbed-end actin filament capping
          actin filament bundle assembly
          actin binding
          structural molecule activity
          structural constituent of cytoskeleton
          calmodulin binding
          spectrin binding
          protein homodimerization activity
          protein heterodimerization activity
          actin filament binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P481381 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P299598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722888 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 426544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927940 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382260 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110485 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35211 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82128 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132941 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101318 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130726 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92116 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P299598 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P481381 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P481381 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P299598 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P481381 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P299598 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P481381 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36537 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89007 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50649 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10856941 - -
          Affymetrix Rat Gene 1.0 ST Array 10856943 - -
          Affymetrix Rat Gene 1.0 ST Array 10856932 - -
          Affymetrix Rat Gene 1.0 ST Array 10856944 - -
          Affymetrix Rat Gene 1.0 ST Array 10856939 - -
          Affymetrix Rat Gene 1.0 ST Array 10856937 - -
          Affymetrix Rat Gene 1.0 ST Array 10856930 - -
          Affymetrix Rat Gene 1.0 ST Array 10856947 - -
          Affymetrix Rat Gene 1.0 ST Array 10856940 - -
          Affymetrix Rat Gene 1.0 ST Array 10856934 - -
          Affymetrix Rat Gene 1.0 ST Array 10856938 - -
          Affymetrix Rat Gene 1.0 ST Array 10856933 - -
          Affymetrix Rat Gene 1.0 ST Array 10856936 - -
          Affymetrix Rat Gene 1.0 ST Array 10856935 - -
          Affymetrix Rat Gene 1.0 ST Array 10856942 - -
          Affymetrix Rat Gene 1.0 ST Array 10856931 - -
          Affymetrix Rat Gene 1.0 ST Array 10856929 - -
          Affymetrix Rat Gene 1.0 ST Array 10856946 - -
          Affymetrix Rat Gene 1.1 ST Array 10856928 - -
          Rat Genome 230 2.0 Array 1392817_at - -
          Rat Genome 230 2.0 Array 1381860_at - -
          Rat Genome 230 2.0 Array 1368603_PM_at - -
          Rat Genome 230 2.0 Array 1368603_at - -
          Rat Genome 230 2.0 Array 1392817_PM_at - -
          Rat Genome 230 2.0 Array 1381860_PM_at - -
          Affymetrix HT Rat Focus Array 1368603_at - -
          Affymetrix HT Rat Focus Array 1368603_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400009248.rn.1 TC0400001305.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400009247.rn.1 TC0400001305.rn.1 RTA TC
          Rat Expression Set 230 1368603_PM_at - -
          Rat Expression Set 230 1368603_at - -
          Affymetrix Rat Gene 1.0 ST Array 10856928 - -
          Rat Genome U34 Set rc_AI136118_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21330271 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350628 - -
          RatRef-12 Expression BeadChip ILMN_62547 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline