Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00562160_m1
          See other TBXAS1 GE Assays ›
          Gene Symbol:
          Tbxas1
          Gene Name:
          thromboxane A synthase 1, platelet
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 66624181 - 66846745 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00562160_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tbxas1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012687.1 NP_036819.1 8 - 9 954 58
          XM_008762759.1 XP_008760981.1 8 - 9 1015 58
          XM_008762760.1 XP_008760982.1 6 - 7 2412 58
          XM_008762761.1 XP_008760983.1 8 - 9 1005 58
          GenBank mRNA D28773.1 - 8 - 9 954 58
          D31798.1 - 8 - 9 876 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400000673.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus Rat Inflammation 1700 Inventoried Rat_Inflammation Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tbxas1

          Panther Classification:

          Molecular Function -

          isomerase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          prostaglandin biosynthetic process
          cellular chloride ion homeostasis
          positive regulation of vasoconstriction
          oxidation-reduction process
          molecular_function
          monooxygenase activity
          thromboxane-A synthase activity
          iron ion binding
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen
          heme binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P260072 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 762079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 670512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 528165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138513 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373627 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517494 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820072 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 260818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633871 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576108 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283546 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37642 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143172 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26666 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88416 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25320 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132666 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37695 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141706 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162038 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153995 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131144 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158899 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93586 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116250 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 123610 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78882 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134755 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176437 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49934 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107270 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79822 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P260072 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P260072 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P260072 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P260072 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700175411 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47125 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54333 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75446 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95847 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17803 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25831 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13972 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31975 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20163 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78552 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77337 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 59925 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87030 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92602 - -
          Agilent Rat cDNA Microarray (G4105A) 700175411 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10854654 - -
          Affymetrix Rat Gene 1.0 ST Array 10854647 - -
          Affymetrix Rat Gene 1.0 ST Array 10854659 - -
          Affymetrix Rat Gene 1.0 ST Array 10854648 - -
          Affymetrix Rat Gene 1.0 ST Array 10854657 - -
          Affymetrix Rat Gene 1.0 ST Array 10854652 - -
          Affymetrix Rat Gene 1.0 ST Array 10854651 - -
          Affymetrix Rat Gene 1.0 ST Array 10854658 - -
          Affymetrix Rat Gene 1.0 ST Array 10854649 - -
          Affymetrix Rat Gene 1.0 ST Array 10854655 - -
          Affymetrix Rat Gene 1.0 ST Array 10854653 - -
          Affymetrix Rat Gene 1.0 ST Array 10854646 - -
          Affymetrix Rat Gene 1.0 ST Array 10854650 - -
          Affymetrix Rat Gene 1.0 ST Array 10854656 - -
          Affymetrix Rat Gene 1.1 ST Array 10854645 - -
          Rat Genome 230 2.0 Array 1368027_PM_at - -
          Rat Genome 230 2.0 Array 1368027_at - -
          Affymetrix HT Rat Focus Array 1368027_PM_at - -
          Affymetrix HT Rat Focus Array 1368027_at - -
          Affymetrix Rat Clariom S Assay PSR0400004909.rn.1 TC0400000673.rn.1 RTA TC
          Rat Expression Set 230 1368027_at - -
          Rat Expression Set 230 1368027_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10854645 - -
          Rat Genome U34 Set D28773_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21725832 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1348927 - -
          RatRef-12 Expression BeadChip ILMN_61548 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline