Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00562334_m1
          See other CALCRL GE Assays ›
          Gene Symbol:
          Calcrl
          Gene Name:
          calcitonin receptor-like
          Gene Aliases:
          CLR, Crlr, RATCRLR, RNCLR
          Chromosome Location:
          Chr.3: 71747940 - 71845487 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00562334_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Calcrl
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012717.1 NP_036849.1 10 - 11 1031 115
          XM_006234438.2 XP_006234500.1 10 - 11 1286 115
          XM_006234439.2 XP_006234501.1 9 - 10 1124 115
          GenBank mRNA L27487.1 - 10 - 11 1304 115
          X70658.1 - 10 - 11 1031 115

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300003427.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Calcrl

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          cAMP biosynthetic process
          calcium ion transport
          G-protein coupled receptor signaling pathway
          adenylate cyclase-activating G-protein coupled receptor signaling pathway
          heart development
          positive regulation of cell proliferation
          protein transport
          positive regulation of cAMP biosynthetic process
          receptor internalization
          negative regulation of smooth muscle contraction
          positive regulation of smooth muscle cell proliferation
          negative regulation of inflammatory response
          cellular response to sucrose stimulus
          adrenomedullin receptor activity
          calcitonin gene-related peptide receptor activity
          calcitonin receptor activity
          protein transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P154358 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P11529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 726364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415634 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313463 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 33066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794776 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97241 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 322431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267455 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143209 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178307 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91339 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37221 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125983 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156517 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20138 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36074 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20731 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126434 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57485 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126226 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11529 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11529 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P11529 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11529 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700937706 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37263 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46495 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94026 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36966 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53777 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104542 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98245 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99729 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50946 - -
          Agilent Rat cDNA Microarray (G4105A) 700937706 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10846766 - -
          Affymetrix Rat Gene 1.0 ST Array 10846769 - -
          Affymetrix Rat Gene 1.0 ST Array 10846774 - -
          Affymetrix Rat Gene 1.0 ST Array 10846772 - -
          Affymetrix Rat Gene 1.0 ST Array 10846777 - -
          Affymetrix Rat Gene 1.0 ST Array 10846770 - -
          Affymetrix Rat Gene 1.0 ST Array 10846776 - -
          Affymetrix Rat Gene 1.0 ST Array 10846778 - -
          Affymetrix Rat Gene 1.0 ST Array 10846768 - -
          Affymetrix Rat Gene 1.0 ST Array 10846779 - -
          Affymetrix Rat Gene 1.0 ST Array 10846775 - -
          Affymetrix Rat Gene 1.0 ST Array 10846771 - -
          Affymetrix Rat Gene 1.0 ST Array 10846780 - -
          Affymetrix Rat Gene 1.0 ST Array 10846773 - -
          Affymetrix Rat Gene 1.0 ST Array 10846764 - -
          Affymetrix Rat Gene 1.0 ST Array 10846763 - -
          Affymetrix Rat Gene 1.0 ST Array 10846765 - -
          Affymetrix Rat Gene 1.0 ST Array 10846767 - -
          Affymetrix Rat Gene 1.1 ST Array 10846762 - -
          Rat Genome 230 2.0 Array 1369647_PM_at - -
          Rat Genome 230 2.0 Array 1377640_at - -
          Rat Genome 230 2.0 Array 1369648_PM_at - -
          Rat Genome 230 2.0 Array 1369648_at - -
          Rat Genome 230 2.0 Array 1369647_at - -
          Rat Genome 230 2.0 Array 1377640_PM_at - -
          Affymetrix HT Rat Focus Array 1369648_at - -
          Affymetrix HT Rat Focus Array 1369647_at - -
          Affymetrix HT Rat Focus Array 1369647_PM_at - -
          Affymetrix HT Rat Focus Array 1369648_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300026916.rn.1 TC0300003427.rn.1 RTA TC
          Rat Expression Set 230 1369648_PM_at - -
          Rat Expression Set 230 1369648_at - -
          Rat Expression Set 230 1369647_at - -
          Rat Expression Set 230 1369647_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10846762 - -
          Rat Genome U34 Set L27487_at - -
          Rat Genome U34 Set rc_AI228025_s_at - -
          Rat Genome U34 Set rc_AI060121_s_at - -
          Rat Genome U34 Set rc_AI060121_s_at - -
          Rat Genome U34 Set L27487_at - -
          Rat Genome U34 Set rc_AI228025_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22303683 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365317 - -
          RatRef-12 Expression BeadChip ILMN_53806 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline