Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00562457_m1
          See other HK2 GE Assays ›
          Gene Symbol:
          Hk2
          Gene Name:
          hexokinase 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 113559396 - 113609897 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00562457_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hk2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012735.2 NP_036867.1 17 - 18 2804 69
          XM_006236711.2 XP_006236773.1 16 - 17 3873 69
          GenBank mRNA AF027179.1 - 17 - 18 2608 69
          M68971.1 - 17 - 18 2805 69
          M68972.1 - 9 - 10 1406 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400003158.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hk2

          Panther Classification:

          Molecular Function -

          kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          cellular glucose homeostasis
          response to ischemia
          glucose metabolic process
          glucose catabolic process
          glycolytic process
          lactation
          apoptotic mitochondrial changes
          regulation of glucose import
          carbohydrate phosphorylation
          glucose 6-phosphate metabolic process
          glucokinase activity
          hexokinase activity
          ATP binding
          glucose binding
          fructokinase activity
          mannokinase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P032574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 33636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412551 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710622 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624098 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527328 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124450 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100835 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22525 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 180786 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25425 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170459 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P032574 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P032574 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P032574 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P452291 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13233 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 657 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6736 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3947 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10863440 - -
          Affymetrix Rat Gene 1.0 ST Array 10863443 - -
          Affymetrix Rat Gene 1.0 ST Array 10863431 - -
          Affymetrix Rat Gene 1.0 ST Array 10863442 - -
          Affymetrix Rat Gene 1.0 ST Array 10863446 - -
          Affymetrix Rat Gene 1.0 ST Array 10863448 - -
          Affymetrix Rat Gene 1.0 ST Array 10863438 - -
          Affymetrix Rat Gene 1.0 ST Array 10863432 - -
          Affymetrix Rat Gene 1.0 ST Array 10863439 - -
          Affymetrix Rat Gene 1.0 ST Array 10863435 - -
          Affymetrix Rat Gene 1.0 ST Array 10863449 - -
          Affymetrix Rat Gene 1.0 ST Array 10863433 - -
          Affymetrix Rat Gene 1.0 ST Array 10863441 - -
          Affymetrix Rat Gene 1.0 ST Array 10863447 - -
          Affymetrix Rat Gene 1.0 ST Array 10863437 - -
          Affymetrix Rat Gene 1.0 ST Array 10863434 - -
          Affymetrix Rat Gene 1.0 ST Array 10863436 - -
          Affymetrix Rat Gene 1.0 ST Array 10863445 - -
          Affymetrix Rat Gene 1.0 ST Array 10863450 - -
          Affymetrix Rat Gene 1.0 ST Array 10863444 - -
          Affymetrix Rat Gene 1.1 ST Array 10863430 - -
          Rat Genome 230 2.0 Array 1383519_at - -
          Rat Genome 230 2.0 Array 1369006_at - -
          Rat Genome 230 2.0 Array 1369006_PM_at - -
          Rat Genome 230 2.0 Array 1383519_PM_at - -
          Affymetrix HT Rat Focus Array 1383519_PM_at - -
          Affymetrix HT Rat Focus Array 1369006_PM_at - -
          Affymetrix HT Rat Focus Array 1383519_at - -
          Affymetrix HT Rat Focus Array 1369006_at - -
          Affymetrix Rat Clariom S Assay PSR0400022283.rn.1 TC0400003158.rn.1 RTA TC
          Rat Expression Set 230 1383519_at - -
          Rat Expression Set 230 1369006_at - -
          Rat Expression Set 230 1369006_PM_at - -
          Rat Expression Set 230 1383519_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10863430 - -
          Rat Genome U34 Set D26393exon_s_at - -
          Rat Genome U34 Set M68971_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21661580 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351347 - -
          RatRef-12 Expression BeadChip ILMN_68744 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline