Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00562543_m1
          See other PCSK2 GE Assays ›
          Gene Symbol:
          Pcsk2
          Gene Name:
          proprotein convertase subtilisin/kexin type 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 137618891 - 137923385 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00562543_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pcsk2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012746.1 NP_036878.1 11 - 12 1752 93
          GenBank mRNA M76706.1 - 11 - 12 1752 93
          M83746.1 - 11 - 12 1727 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300001873.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Protease 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pcsk2

          Panther Classification:

          Molecular Function -

          serine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          nervous system development
          protein processing
          peptide hormone processing
          protein autoprocessing
          insulin processing
          enkephalin processing
          islet amyloid polypeptide processing
          endopeptidase activity
          serine-type endopeptidase activity
          protein binding
          protein complex binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P482629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573916 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167871 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 635174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 420457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732181 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 707309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 651195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661235 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889320 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84095 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442622 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556775 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898463 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 420243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 611390 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 11174 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60440 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 29120 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61613 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95677 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125736 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119687 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161497 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25196 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19894 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107885 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106938 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138141 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132763 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85633 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107938 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176614 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48883 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174635 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 159547 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57008 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59001 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158278 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59252 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14813 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145062 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95210 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 112792 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 173534 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P482629 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P482629 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P482629 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P482629 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P395764 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702151871 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5305 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48647 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69981 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20780 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75162 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77948 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100166 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57294 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31214 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16808 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23055 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79561 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82563 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81431 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93306 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84871 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84732 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76354 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77975 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53567 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46618 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16313 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30044 - -
          Agilent Rat cDNA Microarray (G4105A) 702151871 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10840336 - -
          Affymetrix Rat Gene 1.0 ST Array 10840335 - -
          Affymetrix Rat Gene 1.0 ST Array 10840337 - -
          Affymetrix Rat Gene 1.0 ST Array 10840341 - -
          Affymetrix Rat Gene 1.0 ST Array 10840342 - -
          Affymetrix Rat Gene 1.0 ST Array 10840343 - -
          Affymetrix Rat Gene 1.0 ST Array 10840333 - -
          Affymetrix Rat Gene 1.0 ST Array 10840344 - -
          Affymetrix Rat Gene 1.0 ST Array 10840338 - -
          Affymetrix Rat Gene 1.0 ST Array 10840334 - -
          Affymetrix Rat Gene 1.0 ST Array 10840339 - -
          Affymetrix Rat Gene 1.0 ST Array 10840340 - -
          Affymetrix Rat Gene 1.1 ST Array 10840332 - -
          Rat Genome 230 2.0 Array 1387155_at - -
          Rat Genome 230 2.0 Array 1387155_PM_at - -
          Affymetrix HT Rat Focus Array 1387155_at - -
          Affymetrix HT Rat Focus Array 1387155_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300013407.rn.1 TC0300001873.rn.1 RTA TC
          Rat Expression Set 230 1387155_at - -
          Rat Expression Set 230 1387155_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10840332 - -
          Rat Genome U34 Set M83746_at - -
          Rat Genome U34 Set S40669_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21929985 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370576 - -
          RatRef-12 Expression BeadChip ILMN_65205 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline