Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00562610_m1
          See other ESR2 GE Assays ›
          Gene Symbol:
          Esr2
          Gene Name:
          estrogen receptor 2 (ER beta)
          Gene Aliases:
          ER-beta, ERbeta, Erb2
          Chromosome Location:
          Chr.6: 99164325 - 99214358 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00562610_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (1)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Esr2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012754.1 NP_036886.1 5 - 6 1232 89
          XM_006240221.2 XP_006240283.1 5 - 6 1473 89
          GenBank mRNA AB012721.1 - 4 - 5 816 89
          AB190769.1 - 4 - 5 1055 89
          AB190770.1 - 4 - 5 1055 89
          AF042058.1 - 4 - 5 816 89
          AF042059.1 - 4 - 5 816 89
          AF042060.1 - 3 - 4 699 89
          AF042061.1 - 3 - 4 699 89
          U57439.1 - 5 - 6 1232 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600002686.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Nuclear Receptor Recommended Assay Rattus norvegicus Rat Androgen Panel Rat Estrogen Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Esr2

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          ovarian follicle development
          behavioral fear response
          neuron migration
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          brain development
          learning or memory
          negative regulation of cell proliferation
          male gonad development
          female gonad development
          hormone-mediated apoptotic signaling pathway
          response to water deprivation
          response to hormone
          response to organic cyclic compound
          response to activity
          response to insecticide
          cerebellum development
          amygdala development
          hypothalamus development
          intracellular steroid hormone receptor signaling pathway
          intracellular estrogen receptor signaling pathway
          prostate gland development
          response to nutrient levels
          response to estradiol
          response to testosterone
          response to genistein
          cellular response to drug
          regulation of cell proliferation
          response to drug
          positive regulation of apoptotic process
          steroid hormone mediated signaling pathway
          regulation of neuron apoptotic process
          response to estrogen
          response to human chorionic gonadotropin
          estrous cycle
          response to ethanol
          positive regulation of epidermal growth factor receptor signaling pathway
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of behavior
          negative regulation of smooth muscle cell proliferation
          negative regulation of epithelial cell proliferation
          positive regulation of sequence-specific DNA binding transcription factor activity
          Sertoli cell development
          Sertoli cell proliferation
          uterus development
          vagina development
          negative regulation of cell death
          prostate gland epithelium morphogenesis
          epithelial cell maturation involved in prostate gland development
          negative regulation of androgen receptor signaling pathway
          positive regulation of ERK1 and ERK2 cascade
          cellular response to magnetism
          cellular response to estradiol stimulus
          cellular response to organic cyclic compound
          response to dexamethasone
          negative regulation of neuron death
          response to salt
          response to bisphenol A
          negative regulation of feeding behavior
          negative regulation of reactive oxygen species metabolic process
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          core promoter sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          steroid hormone receptor activity
          RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
          steroid binding
          protein binding
          drug binding
          zinc ion binding
          enzyme binding
          estrogen receptor activity
          estrogen response element binding
          hormone binding
          peroxisome proliferator activated receptor binding
          sequence-specific DNA binding
          estradiol binding
          steroid hormone binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P049530 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P055961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 842710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 578575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 528873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615761 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109515 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66531 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 177537 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124395 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103269 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 123712 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92539 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P049530 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P055960 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P055962 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P049530 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P055962 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P055960 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P055960 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P049530 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P055962 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P442569 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47322 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83936 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41882 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57184 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47631 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77333 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10890659 - -
          Affymetrix Rat Gene 1.0 ST Array 10890666 - -
          Affymetrix Rat Gene 1.0 ST Array 10890668 - -
          Affymetrix Rat Gene 1.0 ST Array 10890664 - -
          Affymetrix Rat Gene 1.0 ST Array 10890657 - -
          Affymetrix Rat Gene 1.0 ST Array 10890665 - -
          Affymetrix Rat Gene 1.0 ST Array 10890663 - -
          Affymetrix Rat Gene 1.0 ST Array 10890658 - -
          Affymetrix Rat Gene 1.0 ST Array 10890669 - -
          Affymetrix Rat Gene 1.0 ST Array 10890670 - -
          Affymetrix Rat Gene 1.0 ST Array 10890667 - -
          Affymetrix Rat Gene 1.0 ST Array 10890661 - -
          Affymetrix Rat Gene 1.0 ST Array 10890660 - -
          Affymetrix Rat Gene 1.0 ST Array 10890656 - -
          Affymetrix Rat Gene 1.1 ST Array 10890654 - -
          Rat Genome 230 2.0 Array 1369760_PM_a_at - -
          Rat Genome 230 2.0 Array 1369760_a_at - -
          Affymetrix HT Rat Focus Array 1369760_a_at - -
          Affymetrix HT Rat Focus Array 1369760_PM_a_at - -
          Affymetrix Rat Clariom S Assay PSR0600019089.rn.1 TC0600002686.rn.1 RTA TC
          Rat Expression Set 230 1369760_PM_a_at - -
          Rat Expression Set 230 1369760_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10890654 - -
          Rat Genome U34 Set U57439_at - -
          Rat Genome U34 Set U57439_g_at - -
          Rat Genome U34 Set U57439_at - -
          Rat Genome U34 Set U57439_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21927223 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354998 - -
          RatRef-12 Expression BeadChip ILMN_64780 - -

          Back To Top
           

          Citations (1)

          Histaminergic responses by hypothalamic neurons that regulate lordosis and their modulation by estradiol.
          Authors
          Dupre C, Lovett-Barron M, Pfaff DW, Kow LM
          Journal
          Proc Natl Acad Sci U S A - Page No 12311 (Tue Jul 06 00:00:00 EDT 2010)
          Volume (Year)
          107 (2010)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline