Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00562673_s1
          See other MAS1 GE Assays ›
          Gene Symbol:
          Mas1
          Gene Name:
          MAS1 proto-oncogene, G protein-coupled receptor
          Gene Aliases:
          Mgra, c-mas
          Chromosome Location:
          Chr.1: 48076768 - 48108218 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00562673_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mas1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012757.2 NP_036889.1 4 1063 67
          XM_006227859.2 XP_006227921.1 3 2325 67
          XM_006227860.2 XP_006227922.1 3 2447 67
          XM_006227867.2 XP_006227929.1 2 2136 67
          XM_006227868.2 XP_006227930.1 2 2084 67
          XM_008758726.1 XP_008756948.1 3 2444 67
          XM_008758727.1 XP_008756949.1 3 2395 67
          XM_008758728.1 XP_008756950.1 3 2091 67
          XM_008758729.1 XP_008756951.1 5 1380 67
          XM_008758730.1 XP_008756952.1 3 2392 67
          GenBank mRNA BC078884.1 - 4 1063 7131760 67

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mas1

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of protein phosphorylation
          G-protein coupled receptor signaling pathway
          activation of NF-kappaB-inducing kinase activity
          spermatogenesis
          positive regulation of cell proliferation
          male gonad development
          response to activity
          hippocampus development
          response to gonadotropin
          angiotensin-activated signaling pathway
          response to drug
          response to peptide hormone
          positive regulation of DNA replication
          regulation of inflammatory response
          positive regulation of inositol phosphate biosynthetic process
          protein kinase C signaling
          cellular response to peptide hormone stimulus
          angiotensin receptor activity
          G-protein coupled receptor activity
          peptide hormone binding
          peptide binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1029253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 815460 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660640 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41329 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57160 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124945 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71784 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1029253 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1029253 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1029253 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1029253 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33945 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20195 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10027 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75534 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10702993 - -
          Affymetrix Rat Gene 1.0 ST Array 10702992 - -
          Affymetrix Rat Gene 1.0 ST Array 10702994 - -
          Affymetrix Rat Gene 1.0 ST Array 10702995 - -
          Affymetrix Rat Gene 1.1 ST Array 10702991 - -
          Rat Genome 230 2.0 Array 1370092_at - -
          Rat Genome 230 2.0 Array 1370092_PM_at - -
          Affymetrix HT Rat Focus Array 1370092_PM_at - -
          Affymetrix HT Rat Focus Array 1370092_at - -
          Affymetrix Rat Clariom S Assay PSR0100003224.rn.1 TC0100000507.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100003215.rn.1 TC0100000507.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100003223.rn.1 TC0100000507.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100003214.rn.1 TC0100000507.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100003222.rn.1 TC0100000507.rn.1 RTA TC
          Rat Expression Set 230 1370092_at - -
          Rat Expression Set 230 1370092_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10702991 - -
          Rat Genome U34 Set rc_AI071498_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22010557 - -
          ABI Rat Genome Survey Microarray 22159430 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350238 - -
          RatRef-12 Expression BeadChip ILMN_68464 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline