Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00563433_m1
          See other KCNC1 GE Assays ›
          Gene Symbol:
          Kcnc1
          Gene Name:
          potassium channel, voltage gated Shaw-related subfamily C, member 1
          Gene Aliases:
          KShIIIB, Kv3.1, Kv4, NGK2-KV4
          Chromosome Location:
          Chr.1: 102414625 - 102456411 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00563433_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kcnc1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012856.1 NP_036988.1 2 - 3 2666 67
          GenBank mRNA M68880.1 - 2 - 3 2666 67
          X62840.1 - 2 - 3 1791 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100001524.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Kcnc1

          Panther Classification:

          Molecular Function -

          voltage-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          brain development
          response to toxic substance
          response to light intensity
          response to auditory stimulus
          response to amine
          cerebellum development
          globus pallidus development
          regulation of ion transmembrane transport
          positive regulation of ion transmembrane transport
          cellular response to drug
          response to potassium ion
          protein homooligomerization
          response to fibroblast growth factor
          potassium ion transmembrane transport
          regulation of potassium ion transmembrane transport
          positive regulation of potassium ion transmembrane transport
          response to nerve growth factor
          voltage-gated potassium channel activity
          delayed rectifier potassium channel activity
          protein binding
          kinesin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P426079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 420679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513360 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166964 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33798 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135271 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5185 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109901 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63945 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69209 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P426079 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P426079 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P426079 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P426079 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701914110 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48082 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74911 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77679 - -
          Agilent Rat cDNA Microarray (G4105A) 701914110 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10707117 - -
          Affymetrix Rat Gene 1.0 ST Array 10707116 - -
          Affymetrix Rat Gene 1.0 ST Array 10707114 - -
          Affymetrix Rat Gene 1.0 ST Array 10707115 - -
          Affymetrix Rat Gene 1.1 ST Array 10707113 - -
          Rat Genome 230 2.0 Array 1368524_at - -
          Rat Genome 230 2.0 Array 1368524_PM_at - -
          Affymetrix HT Rat Focus Array 1368524_PM_at - -
          Affymetrix HT Rat Focus Array 1368524_at - -
          Affymetrix Rat Clariom S Assay PSR0100012313.rn.1 TC0100001524.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100012311.rn.1 TC0100001524.rn.1 RTA TC
          Rat Expression Set 230 1368524_at - -
          Rat Expression Set 230 1368524_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10707113 - -
          Rat Genome U34 Set X62840mRNA_s_at - -
          Rat Genome U34 Set X62840mRNA_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21407457 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_2040259 - -
          RatRef-12 Expression BeadChip ILMN_62192 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline