Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00563487_m1
          See other NFYC GE Assays ›
          Gene Symbol:
          Nfyc
          Gene Name:
          nuclear transcription factor-Y gamma
          Gene Aliases:
          CBF-C
          Chromosome Location:
          Chr.5: 139687442 - 139759741 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00563487_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nfyc
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012866.2 NP_036998.1 7 - 8 918 58
          XM_006238785.2 XP_006238847.1 7 - 8 891 58
          GenBank mRNA BC093619.1 - 7 - 8 918 7314492 58
          U17607.1 - 7 - 8 806 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500003522.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nfyc

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          transcription factor binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P400520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898095 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873691 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 762386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244125 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 102980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345691 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 387436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 386596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95127 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178085 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124027 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108838 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81160 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3114 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71798 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17091 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102031 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P400520 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P400520 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P400520 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P400520 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P174682 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700420281 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62026 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36883 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39721 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67018 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73586 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99184 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34642 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25222 - -
          Agilent Rat cDNA Microarray (G4105A) 700420281 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10879434 - -
          Affymetrix Rat Gene 1.0 ST Array 10879439 - -
          Affymetrix Rat Gene 1.0 ST Array 10879438 - -
          Affymetrix Rat Gene 1.0 ST Array 10879437 - -
          Affymetrix Rat Gene 1.0 ST Array 10879441 - -
          Affymetrix Rat Gene 1.0 ST Array 10879433 - -
          Affymetrix Rat Gene 1.0 ST Array 10879431 - -
          Affymetrix Rat Gene 1.0 ST Array 10879436 - -
          Affymetrix Rat Gene 1.0 ST Array 10879429 - -
          Affymetrix Rat Gene 1.0 ST Array 10879428 - -
          Affymetrix Rat Gene 1.0 ST Array 10879435 - -
          Affymetrix Rat Gene 1.1 ST Array 10879427 - -
          Rat Genome 230 2.0 Array 1398268_PM_at - -
          Rat Genome 230 2.0 Array 1398268_at - -
          Rat Genome 230 2.0 Array 1368820_PM_at - -
          Rat Genome 230 2.0 Array 1368820_at - -
          Affymetrix HT Rat Focus Array 1368820_at - -
          Affymetrix HT Rat Focus Array 1398268_at - -
          Affymetrix HT Rat Focus Array 1368820_PM_at - -
          Affymetrix HT Rat Focus Array 1398268_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500026085.rn.1 TC0500003522.rn.1 RTA TC
          Rat Expression Set 230 1368820_at - -
          Rat Expression Set 230 1398268_PM_at - -
          Rat Expression Set 230 1398268_at - -
          Rat Expression Set 230 1368820_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10879427 - -
          Rat Genome U34 Set U17607_at - -
          Rat Genome U34 Set rc_AA875121_at - -
          Rat Genome U34 Set U17607_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20944425 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358630 - -
          RatRef-12 Expression BeadChip ILMN_64400 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline