Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00563688_m1
          See other ADRBK2 GE Assays ›
          Gene Symbol:
          Adrbk2
          Gene Name:
          adrenergic, beta, receptor kinase 2
          Gene Aliases:
          Grk3
          Chromosome Location:
          Chr.12: 49626871 - 49750389 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00563688_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Adrbk2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012897.2 NP_037029.1 19 - 20 1794 128
          XM_008769317.1 XP_008767539.1 18 - 19 1685 128

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200000845.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Adrbk2

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          desensitization of G-protein coupled receptor protein signaling pathway
          protein phosphorylation
          intracellular protein transport
          signal transduction
          multicellular organism aging
          receptor internalization
          termination of G-protein coupled receptor signaling pathway
          inositol phosphate metabolic process
          negative regulation of G-protein coupled receptor protein signaling pathway
          rhodopsin metabolic process
          protein serine/threonine kinase activity
          G-protein coupled receptor kinase activity
          ATP binding
          D1 dopamine receptor binding
          beta-adrenergic receptor kinase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P146640 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P146640 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P146640 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P146640 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P146640 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103931 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40752 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66249 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23526 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 59453 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85098 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 59476 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 59099 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10931627 - -
          Affymetrix Rat Gene 1.0 ST Array 10931623 - -
          Affymetrix Rat Gene 1.0 ST Array 10931621 - -
          Affymetrix Rat Gene 1.0 ST Array 10931629 - -
          Affymetrix Rat Gene 1.0 ST Array 10931636 - -
          Affymetrix Rat Gene 1.0 ST Array 10931625 - -
          Affymetrix Rat Gene 1.0 ST Array 10931637 - -
          Affymetrix Rat Gene 1.0 ST Array 10931630 - -
          Affymetrix Rat Gene 1.0 ST Array 10931634 - -
          Affymetrix Rat Gene 1.0 ST Array 10931626 - -
          Affymetrix Rat Gene 1.0 ST Array 10931624 - -
          Affymetrix Rat Gene 1.0 ST Array 10931633 - -
          Affymetrix Rat Gene 1.0 ST Array 10931622 - -
          Affymetrix Rat Gene 1.0 ST Array 10931631 - -
          Affymetrix Rat Gene 1.0 ST Array 10931628 - -
          Affymetrix Rat Gene 1.0 ST Array 10931632 - -
          Affymetrix Rat Gene 1.0 ST Array 10931635 - -
          Affymetrix Rat Gene 1.1 ST Array 10931620 - -
          Rat Genome 230 2.0 Array 1381361_at - -
          Rat Genome 230 2.0 Array 1369403_PM_at - -
          Rat Genome 230 2.0 Array 1369403_at - -
          Rat Genome 230 2.0 Array 1381361_PM_at - -
          Affymetrix HT Rat Focus Array 1369403_at - -
          Affymetrix HT Rat Focus Array 1369403_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1200007414.rn.1 TC1200000845.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200007415.rn.1 TC1200000845.rn.1 RTA TC
          Rat Expression Set 230 1369403_PM_at - -
          Rat Expression Set 230 1369403_at - -
          Affymetrix Rat Gene 1.0 ST Array 10931620 - -
          Rat Genome U34 Set M87855_at - -
          Rat Genome U34 Set M87855_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21419287 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376414 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline