Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00564227_m1
          See other ICAM1 GE Assays ›
          Gene Symbol:
          Icam1
          Gene Name:
          intercellular adhesion molecule 1
          Gene Aliases:
          CD54, ICAM
          Chromosome Location:
          Chr.8: 22035287 - 22047049 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00564227_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Icam1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012967.1 NP_037099.1 1 - 2 103 61
          GenBank mRNA BC081837.1 - 1 - 2 115 7122397 61
          D00913.1 - 1 - 2 103 61
          FQ228241.1 - 1 - 2 123 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800000214.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Rat Inflammation 1700 Inventoried Rat_Inflammation Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Icam1

          Gene Ontology Categories:

          Function(s) Process(es)

          ovarian follicle development
          response to hypoxia
          response to amphetamine
          T cell activation via T cell receptor contact with antigen bound to MHC molecule on antigen presenting cell
          acute inflammatory response to antigenic stimulus
          T cell antigen processing and presentation
          positive regulation of cellular extravasation
          cell adhesion
          leukocyte cell-cell adhesion
          cell aging
          sensory perception of sound
          regulation of cell shape
          response to ionizing radiation
          response to sulfur dioxide
          response to organic cyclic compound
          single organismal cell-cell adhesion
          membrane to membrane docking
          regulation of cell adhesion
          positive regulation of actin filament polymerization
          cellular response to nutrient levels
          response to lipopolysaccharide
          cell adhesion mediated by integrin
          response to gonadotropin
          response to drug
          response to amino acid
          positive regulation of GTPase activity
          adhesion of symbiont to host
          positive regulation of nitric oxide biosynthetic process
          response to ethanol
          positive regulation of vasoconstriction
          response to copper ion
          receptor-mediated virion attachment to host cell
          positive regulation of peptidyl-tyrosine phosphorylation
          leukocyte migration
          positive regulation of NF-kappaB transcription factor activity
          negative regulation of calcium ion transport
          establishment of endothelial barrier
          positive regulation of ERK1 and ERK2 cascade
          cellular response to lipopolysaccharide
          cellular response to organic substance
          cellular response to alkaloid
          cellular response to glucose stimulus
          cellular response to interleukin-1
          cellular response to tumor necrosis factor
          cellular response to hypoxia
          establishment of Sertoli cell barrier
          regulation of ruffle assembly
          negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
          negative regulation of endothelial cell apoptotic process
          integrin binding
          protein complex binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P15253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723697 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 1888 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15253 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15253 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P15253 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15253 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12194 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10908321 - -
          Affymetrix Rat Gene 1.0 ST Array 10908324 - -
          Affymetrix Rat Gene 1.0 ST Array 10908323 - -
          Affymetrix Rat Gene 1.0 ST Array 10908325 - -
          Affymetrix Rat Gene 1.0 ST Array 10908322 - -
          Affymetrix Rat Gene 1.0 ST Array 10908320 - -
          Affymetrix Rat Gene 1.0 ST Array 10908327 - -
          Affymetrix Rat Gene 1.0 ST Array 10908326 - -
          Affymetrix Rat Gene 1.1 ST Array 10908319 - -
          Rat Genome 230 2.0 Array 1387202_at - -
          Rat Genome 230 2.0 Array 1387202_PM_at - -
          Affymetrix HT Rat Focus Array 1387202_at - -
          Affymetrix HT Rat Focus Array 1387202_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0800001130.rn.1 TC0800000214.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800001126.rn.1 TC0800000214.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800001127.rn.1 TC0800000214.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800001131.rn.1 TC0800000214.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800001125.rn.1 TC0800000214.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800001132.rn.1 TC0800000214.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800001128.rn.1 TC0800000214.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800001129.rn.1 TC0800000214.rn.1 RTA TC
          Rat Expression Set 230 1387202_at - -
          Rat Expression Set 230 1387202_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10908319 - -
          Rat Genome U34 Set D00913_at - -
          Rat Genome U34 Set D00913_g_at - -
          Rat Genome U34 Set D00913_g_at - -
          Rat Genome U34 Set D00913_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21010672 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354506 - -
          RatRef-12 Expression BeadChip ILMN_61139 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-xpdfw:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline