Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00564454_m1
          See other P2RX1 GE Assays ›
          Gene Symbol:
          P2rx1
          Gene Name:
          purinergic receptor P2X, ligand-gated ion channel, 1
          Gene Aliases:
          P2XMR
          Chromosome Location:
          Chr.10: 59566268 - 59581328 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00564454_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: P2rx1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001142367.2 NP_001135839.1 7 - 8 875 59
          NM_001285415.1 NP_001272344.1 6 - 7 875 59
          NM_012997.3 NP_037129.1 7 - 8 956 59
          GenBank mRNA AF231010.1 - 6 - 7 666 59
          BC061742.1 - 7 - 8 948 5599053 59
          FJ424511.1 - 7 - 8 666 59
          M80602.1 - 1 - 2 183 59
          X80477.1 - 7 - 8 956 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000001018.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene P2rx1

          Panther Classification:

          Molecular Function -

          ligand-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          serotonin secretion by platelet
          regulation of vascular smooth muscle contraction
          ion transport
          cation transport
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          regulation of smooth muscle contraction
          insemination
          regulation of blood pressure
          response to organic substance
          neuronal action potential
          regulation of vasoconstriction
          platelet activation
          response to ATP
          synaptic transmission, glutamatergic
          purinergic nucleotide receptor signaling pathway
          vasoconstriction
          positive regulation of ion transport
          ceramide biosynthetic process
          protein homooligomerization
          protein heterooligomerization
          regulation of calcium ion transport
          cation transmembrane transport
          purinergic nucleotide receptor activity
          extracellular ATP-gated cation channel activity
          cation channel activity
          protein binding
          ATP binding
          drug binding
          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P019265 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P135082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609120 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572312 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32859 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117943 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P019265 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P135082 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P135082 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P019265 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P019265 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P135082 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P135082 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700143373 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48608 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22284 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66892 - -
          Agilent Rat cDNA Microarray (G4105A) 700143373 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10735609 - -
          Affymetrix Rat Gene 1.0 ST Array 10735605 - -
          Affymetrix Rat Gene 1.0 ST Array 10735603 - -
          Affymetrix Rat Gene 1.0 ST Array 10735602 - -
          Affymetrix Rat Gene 1.0 ST Array 10735614 - -
          Affymetrix Rat Gene 1.0 ST Array 10735612 - -
          Affymetrix Rat Gene 1.0 ST Array 10735611 - -
          Affymetrix Rat Gene 1.0 ST Array 10735613 - -
          Affymetrix Rat Gene 1.0 ST Array 10735608 - -
          Affymetrix Rat Gene 1.0 ST Array 10735607 - -
          Affymetrix Rat Gene 1.0 ST Array 10735604 - -
          Affymetrix Rat Gene 1.0 ST Array 10735610 - -
          Affymetrix Rat Gene 1.0 ST Array 10735601 - -
          Affymetrix Rat Gene 1.0 ST Array 10735606 - -
          Affymetrix Rat Gene 1.1 ST Array 10735600 - -
          Rat Genome 230 2.0 Array 1385148_PM_at - -
          Rat Genome 230 2.0 Array 1369178_a_at - -
          Rat Genome 230 2.0 Array 1385148_at - -
          Rat Genome 230 2.0 Array 1369178_PM_a_at - -
          Affymetrix HT Rat Focus Array 1369178_a_at - -
          Affymetrix HT Rat Focus Array 1369178_PM_a_at - -
          Affymetrix HT Rat Focus Array 1385148_PM_at - -
          Affymetrix HT Rat Focus Array 1385148_at - -
          Affymetrix Rat Clariom S Assay PSR1000009271.rn.1 TC1000001018.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000009272.rn.1 TC1000001018.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000009268.rn.1 TC1000001018.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000009270.rn.1 TC1000001018.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000009258.rn.1 TC1000001018.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000009273.rn.1 TC1000001018.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000009264.rn.1 TC1000001018.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000009274.rn.1 TC1000001018.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000009260.rn.1 TC1000001018.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000009261.rn.1 TC1000001018.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000009269.rn.1 TC1000001018.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000009259.rn.1 TC1000001018.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000009257.rn.1 TC1000001018.rn.1 RTA TC
          Rat Expression Set 230 1369178_a_at - -
          Rat Expression Set 230 1369178_PM_a_at - -
          Rat Expression Set 230 1385148_PM_at - -
          Rat Expression Set 230 1385148_at - -
          Affymetrix Rat Gene 1.0 ST Array 10735600 - -
          Rat Genome U34 Set X80477_at - -
          Rat Genome U34 Set X80477_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21083459 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372182 - -
          RatRef-12 Expression BeadChip ILMN_48600 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline