Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00564609_m1
          See other SIRPA GE Assays ›
          Gene Symbol:
          Sirpa
          Gene Name:
          signal-regulatory protein alpha
          Gene Aliases:
          Bit, Ptpns1, SHPS-1
          Chromosome Location:
          Chr.3: 122113384 - 122152478 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00564609_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sirpa
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013016.2 NP_037148.2 3 - 4 680 91
          XM_006234951.2 XP_006235013.1 2 - 3 978 91
          XM_006234952.2 XP_006235014.1 3 - 4 1340 91
          XM_006234953.2 XP_006235015.1 3 - 4 1522 91
          GenBank mRNA AF055065.1 - 3 - 4 544 91
          BC099773.1 - 3 - 4 616 7932298 91
          D38468.1 - 2 - 3 735 91
          D85183.1 - 2 - 3 492 91
          U62328.1 - 3 - 4 680 91

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300001702.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sirpa

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          hematopoietic progenitor cell differentiation
          phagocytosis, recognition
          phagocytosis, engulfment
          cytoskeleton organization
          actin filament organization
          cell-matrix adhesion
          cell migration
          positive regulation of phagocytosis
          protein binding
          SH3 domain binding
          protein phosphorylated amino acid binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P000683 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P000678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 387548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 707639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542691 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564413 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79122 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141610 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33449 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55273 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145047 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P000678 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P000683 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P000683 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P000678 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P000683 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P000678 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P14890 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701297222 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 635 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11310 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8901 - -
          Agilent Rat cDNA Microarray (G4105A) 701297222 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10839799 - -
          Affymetrix Rat Gene 1.0 ST Array 10839797 - -
          Affymetrix Rat Gene 1.0 ST Array 10839800 - -
          Affymetrix Rat Gene 1.0 ST Array 10839795 - -
          Affymetrix Rat Gene 1.0 ST Array 10839798 - -
          Affymetrix Rat Gene 1.0 ST Array 10839802 - -
          Affymetrix Rat Gene 1.0 ST Array 10839794 - -
          Affymetrix Rat Gene 1.0 ST Array 10839792 - -
          Affymetrix Rat Gene 1.0 ST Array 10839793 - -
          Affymetrix Rat Gene 1.0 ST Array 10839796 - -
          Affymetrix Rat Gene 1.0 ST Array 10839801 - -
          Affymetrix Rat Gene 1.1 ST Array 10839790 - -
          Rat Genome 230 2.0 Array 1386057_PM_at - -
          Rat Genome 230 2.0 Array 1370574_PM_a_at - -
          Rat Genome 230 2.0 Array 1386057_at - -
          Rat Genome 230 2.0 Array 1370574_a_at - -
          Rat Genome 230 2.0 Array 1367881_at - -
          Rat Genome 230 2.0 Array 1367881_PM_at - -
          Affymetrix HT Rat Focus Array 1370574_PM_a_at - -
          Affymetrix HT Rat Focus Array 1367881_at - -
          Affymetrix HT Rat Focus Array 1367881_PM_at - -
          Affymetrix HT Rat Focus Array 1370574_a_at - -
          Affymetrix Rat Clariom S Assay PSR0300012180.rn.1 TC0300001702.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300012181.rn.1 TC0300001702.rn.1 RTA TC
          Rat Expression Set 230 1386057_at - -
          Rat Expression Set 230 1370574_PM_a_at - -
          Rat Expression Set 230 1367881_PM_at - -
          Rat Expression Set 230 1370574_a_at - -
          Rat Expression Set 230 1367881_at - -
          Rat Expression Set 230 1386057_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10839790 - -
          Rat Genome U34 Set AF055065_s_at - -
          Rat Genome U34 Set D38468_s_at - -
          Rat Genome U34 Set D85183_s_at - -
          Rat Genome U34 Set rc_AA943810_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21376564 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360759 - -
          RatRef-12 Expression BeadChip ILMN_68010 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline