Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00565033_m1
          See other PPP1CB GE Assays ›
          Gene Symbol:
          Ppp1cb
          Gene Name:
          protein phosphatase 1, catalytic subunit, beta isozyme
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 23548507 - 23581141 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00565033_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ppp1cb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013065.2 NP_037197.1 1 - 2 319 78
          XM_008764505.1 XP_008762727.1 2 - 3 257 78
          GenBank mRNA BC062033.1 - 1 - 2 319 5623630 78
          D90164.1 - 1 - 2 99 78
          FQ212294.1 - 2 - 3 257 78
          S78218.1 - 1 - 2 86 78

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ppp1cb

          Panther Classification:

          Molecular Function -

          protein phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          glycogen metabolic process
          regulation of glycogen biosynthetic process
          regulation of glycogen catabolic process
          protein dephosphorylation
          regulation of cell adhesion
          circadian regulation of gene expression
          regulation of circadian rhythm
          entrainment of circadian clock by photoperiod
          cell division
          phosphoprotein phosphatase activity
          protein serine/threonine phosphatase activity
          protein binding
          phosphatase activity
          myosin phosphatase activity
          protein kinase binding
          metal ion binding
          myosin-light-chain-phosphatase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P11689 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P076765 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P269596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968782 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865055 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20410 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33502 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 38696 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 112263 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156749 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P076765 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P269596 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11689 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P269596 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11689 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P076765 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P269596 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P11689 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P076765 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P279046 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11689 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700369892 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29306 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23471 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15075 - -
          Agilent Rat cDNA Microarray (G4105A) 700369892 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10806493 - -
          Affymetrix Rat Gene 1.0 ST Array 10809903 - -
          Affymetrix Rat Gene 1.0 ST Array 10888693 - -
          Affymetrix Rat Gene 1.0 ST Array 10888698 - -
          Affymetrix Rat Gene 1.0 ST Array 10888695 - -
          Affymetrix Rat Gene 1.0 ST Array 10888694 - -
          Affymetrix Rat Gene 1.0 ST Array 10888699 - -
          Affymetrix Rat Gene 1.0 ST Array 10888696 - -
          Affymetrix Rat Gene 1.0 ST Array 10809881 - -
          Affymetrix Rat Gene 1.0 ST Array 10888700 - -
          Affymetrix Rat Gene 1.0 ST Array 10888697 - -
          Affymetrix Rat Gene 1.1 ST Array 10809902 - -
          Affymetrix Rat Gene 1.1 ST Array 10888692 - -
          Affymetrix Rat Gene 1.1 ST Array 10809880 - -
          Affymetrix Rat Gene 1.1 ST Array 10806492 - -
          Rat Genome 230 2.0 Array 1386950_at - -
          Rat Genome 230 2.0 Array 1386950_PM_at - -
          Rat Genome 230 2.0 Array 1390990_at - -
          Rat Genome 230 2.0 Array 1390990_PM_at - -
          Affymetrix HT Rat Focus Array 1386950_at - -
          Affymetrix HT Rat Focus Array 1386950_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0600014102.rn.1 TC0600001974.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600014107.rn.1 TC0600001974.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600014106.rn.1 TC0600001974.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600014103.rn.1 TC0600001974.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600014108.rn.1 TC0600001974.rn.1 RTA TC
          Rat Expression Set 230 1386950_PM_at - -
          Rat Expression Set 230 1386950_at - -
          Affymetrix Rat Gene 1.0 ST Array 10888692 - -
          Rat Genome U34 Set S78218_at - -
          Rat Genome U34 Set S78218_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21820506 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368894 - -
          RatRef-12 Expression BeadChip ILMN_62999 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline