Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00565331_m1
          See other SMAD3 GE Assays ›
          Gene Symbol:
          Smad3
          Gene Name:
          SMAD family member 3
          Gene Aliases:
          Madh3, Smad 3, mad3
          Chromosome Location:
          Chr.8: 68569530 - 68678349 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00565331_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Smad3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013095.3 NP_037227.1 4 - 5 806 94
          XM_008766216.1 XP_008764438.1 2 - 3 745 94
          XM_008766217.1 XP_008764439.1 2 - 3 471 94
          GenBank mRNA BC064437.1 - 806 5599910 94
          DQ409172.1 - 1 - 2 62 94
          U66479.1 - 4 - 5 615 94

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the XM_008766216.1, XM_008766217.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Smad3

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor gene-specific transcriptional regulator

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          skeletal system development
          osteoblast differentiation
          ureteric bud development
          response to hypoxia
          in utero embryonic development
          mesoderm formation
          somitogenesis
          liver development
          negative regulation of protein phosphorylation
          heart looping
          osteoblast development
          immune system development
          transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transport
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          immune response
          cell cycle arrest
          transforming growth factor beta receptor signaling pathway
          SMAD protein complex assembly
          gastrulation
          endoderm development
          negative regulation of cell proliferation
          embryonic pattern specification
          positive regulation of gene expression
          positive regulation of alkaline phosphatase activity
          positive regulation of epithelial to mesenchymal transition
          regulation of striated muscle tissue development
          regulation of transforming growth factor beta receptor signaling pathway
          evasion or tolerance of host defenses by virus
          signal transduction involved in regulation of gene expression
          negative regulation of cell growth
          positive regulation of cell migration
          positive regulation of bone mineralization
          thyroid gland development
          positive regulation of chondrocyte differentiation
          positive regulation of interleukin-1 beta production
          regulation of transforming growth factor beta2 production
          positive regulation of transforming growth factor beta3 production
          activin receptor signaling pathway
          negative regulation of osteoblast proliferation
          positive regulation of catenin import into nucleus
          intracellular signal transduction
          nodal signaling pathway
          T cell activation
          negative regulation of protein catabolic process
          positive regulation of transcription factor import into nucleus
          negative regulation of apoptotic process
          cell-cell junction organization
          negative regulation of osteoblast differentiation
          positive regulation of transcription, DNA-templated
          negative regulation of mitotic cell cycle
          positive regulation of transcription from RNA polymerase II promoter
          paraxial mesoderm morphogenesis
          developmental growth
          embryonic foregut morphogenesis
          embryonic cranial skeleton morphogenesis
          regulation of epithelial cell proliferation
          negative regulation of inflammatory response
          regulation of immune response
          protein stabilization
          positive regulation of positive chemotaxis
          regulation of binding
          positive regulation of stress fiber assembly
          positive regulation of focal adhesion assembly
          pericardium development
          transdifferentiation
          SMAD protein signal transduction
          negative regulation of wound healing
          lens fiber cell differentiation
          positive regulation of canonical Wnt signaling pathway
          extrinsic apoptotic signaling pathway
          activation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
          positive regulation of extracellular matrix assembly
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcription factor activity, RNA polymerase II core promoter sequence-specific
          core promoter proximal region sequence-specific DNA binding
          RNA polymerase II activating transcription factor binding
          DNA binding
          chromatin binding
          double-stranded DNA binding
          transcription factor activity, sequence-specific DNA binding
          transforming growth factor beta receptor binding
          protein binding
          collagen binding
          beta-catenin binding
          transcription factor binding
          zinc ion binding
          enzyme binding
          protein kinase binding
          phosphatase binding
          transforming growth factor beta receptor, pathway-specific cytoplasmic mediator activity
          chromatin DNA binding
          ubiquitin protein ligase binding
          identical protein binding
          protein homodimerization activity
          ubiquitin binding
          bHLH transcription factor binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          SMAD binding
          protein heterodimerization activity
          co-SMAD binding
          R-SMAD binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P068167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448894 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 651607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 639847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101098 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319489 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580042 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41443 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6643 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120321 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 96270 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111640 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101468 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137341 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115815 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63675 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153782 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130310 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 34141 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12075 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93016 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127986 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P068167 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P068167 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P068167 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P531593 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 59122 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21872 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61087 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88223 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98233 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2458 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74115 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10918184 - -
          Affymetrix Rat Gene 1.0 ST Array 10918179 - -
          Affymetrix Rat Gene 1.0 ST Array 10918185 - -
          Affymetrix Rat Gene 1.0 ST Array 10918180 - -
          Affymetrix Rat Gene 1.0 ST Array 10918183 - -
          Affymetrix Rat Gene 1.0 ST Array 10918172 - -
          Affymetrix Rat Gene 1.0 ST Array 10918182 - -
          Affymetrix Rat Gene 1.0 ST Array 10918170 - -
          Affymetrix Rat Gene 1.0 ST Array 10918178 - -
          Affymetrix Rat Gene 1.0 ST Array 10918171 - -
          Affymetrix Rat Gene 1.0 ST Array 10918175 - -
          Affymetrix Rat Gene 1.0 ST Array 10918174 - -
          Affymetrix Rat Gene 1.0 ST Array 10918177 - -
          Affymetrix Rat Gene 1.0 ST Array 10918176 - -
          Affymetrix Rat Gene 1.0 ST Array 10918181 - -
          Affymetrix Rat Gene 1.0 ST Array 10918173 - -
          Affymetrix Rat Gene 1.1 ST Array 10918169 - -
          Rat Genome 230 2.0 Array 1369528_at - -
          Rat Genome 230 2.0 Array 1369528_PM_at - -
          Rat Genome 230 2.0 Array 1389127_PM_at - -
          Rat Genome 230 2.0 Array 1389127_at - -
          Affymetrix HT Rat Focus Array 1369528_at - -
          Affymetrix HT Rat Focus Array 1369528_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0800022256.rn.1 TC0800002735.rn.1 RTA TC
          Rat Expression Set 230 1369528_at - -
          Rat Expression Set 230 1369528_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10918169 - -
          Rat Genome U34 Set U66479_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21110267 - -
          ABI Rat Genome Survey Microarray 22199973 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354074 - -
          RatRef-12 Expression BeadChip ILMN_59033 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline