Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00565950_m1
          See other TCF12 GE Assays ›
          Gene Symbol:
          Tcf12
          Gene Name:
          transcription factor 12
          Gene Aliases:
          SCBPA
          Chromosome Location:
          Chr.8: 78343634 - 78655856 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00565950_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tcf12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013176.2 NP_037308.1 16 - 17 1672 73
          XM_006243345.1 XP_006243407.1 16 - 17 1527 73
          XM_006243346.1 XP_006243408.1 16 - 17 1527 73
          XM_006243347.1 XP_006243409.1 15 - 16 1458 73
          XM_006243348.1 XP_006243410.1 15 - 16 1455 73
          XM_006243349.1 XP_006243411.1 15 - 16 1455 73
          XM_006243350.1 XP_006243412.1 15 - 16 1452 73
          XM_006243351.1 XP_006243413.1 14 - 15 1383 73
          XM_006243352.2 XP_006243414.1 9 - 10 1321 73
          XM_006243353.2 XP_006243415.1 9 - 10 1318 73
          XM_006243354.2 XP_006243416.1 8 - 9 1246 73
          XM_008766218.1 XP_008764440.1 14 - 15 5211 73
          XM_008766219.1 XP_008764441.1 8 - 9 1249 73
          GenBank mRNA L09656.1 - 16 - 17 1672 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800002886.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tcf12

          Panther Classification:

          Molecular Function -

          basic helix-loop-helix transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          positive regulation of gene expression
          positive regulation of neuron differentiation
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          transcription factor binding
          cAMP response element binding
          bHLH transcription factor binding
          transcription regulatory region DNA binding
          SMAD binding
          protein heterodimerization activity
          protein dimerization activity
          E-box binding
          HMG box domain binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P079935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204494 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 71179 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 228103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 694533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603109 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935299 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890218 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421505 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 372136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 528288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 322903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 126211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641668 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301504 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74807 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 148459 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121224 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71761 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41493 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 159399 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 29363 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126310 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130403 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37558 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10671 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 180773 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60144 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119793 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46825 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142354 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 18900 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81802 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178922 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 9768 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27331 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P079935 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P079935 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P079935 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P362992 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84576 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93595 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83334 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88764 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86621 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86918 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17137 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57523 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8412 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2195 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40781 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26568 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52400 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10918624 - -
          Affymetrix Rat Gene 1.0 ST Array 10918630 - -
          Affymetrix Rat Gene 1.0 ST Array 10918626 - -
          Affymetrix Rat Gene 1.0 ST Array 10918628 - -
          Affymetrix Rat Gene 1.0 ST Array 10918635 - -
          Affymetrix Rat Gene 1.0 ST Array 10918621 - -
          Affymetrix Rat Gene 1.0 ST Array 10918622 - -
          Affymetrix Rat Gene 1.0 ST Array 10918627 - -
          Affymetrix Rat Gene 1.0 ST Array 10918637 - -
          Affymetrix Rat Gene 1.0 ST Array 10918639 - -
          Affymetrix Rat Gene 1.0 ST Array 10918636 - -
          Affymetrix Rat Gene 1.0 ST Array 10918631 - -
          Affymetrix Rat Gene 1.0 ST Array 10918632 - -
          Affymetrix Rat Gene 1.0 ST Array 10918625 - -
          Affymetrix Rat Gene 1.0 ST Array 10918633 - -
          Affymetrix Rat Gene 1.0 ST Array 10918629 - -
          Affymetrix Rat Gene 1.0 ST Array 10918623 - -
          Affymetrix Rat Gene 1.0 ST Array 10918638 - -
          Affymetrix Rat Gene 1.1 ST Array 10918620 - -
          Rat Genome 230 2.0 Array 1387374_PM_at - -
          Rat Genome 230 2.0 Array 1387374_at - -
          Affymetrix HT Rat Focus Array 1387374_PM_at - -
          Affymetrix HT Rat Focus Array 1387374_at - -
          Affymetrix Rat Clariom S Assay PSR0800023352.rn.1 TC0800002886.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800023351.rn.1 TC0800002886.rn.1 RTA TC
          Rat Expression Set 230 1387374_PM_at - -
          Rat Expression Set 230 1387374_at - -
          Affymetrix Rat Gene 1.0 ST Array 10918620 - -
          Rat Genome U34 Set L09656_at - -
          Rat Genome U34 Set S53913_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22390291 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376542 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline