Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00566528_m1
          See other NQO1 GE Assays ›
          Gene Symbol:
          Nqo1
          Gene Name:
          NAD(P)H dehydrogenase, quinone 1
          Gene Aliases:
          Dia4
          Chromosome Location:
          Chr.19: 38422210 - 38437103 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00566528_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nqo1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017000.3 NP_058696.2 5 - 6 607 90
          GenBank mRNA BC083542.1 - 5 - 6 607 7190779 90
          J02608.1 - 5 - 6 592 90
          J02640.1 - 4 - 5 566 90
          J02679.1 - 5 - 6 592 90
          X17464.1 - 3 - 4 281 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900000518.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nqo1

          Panther Classification:

          Molecular Function -

          oxidoreductase

          Gene Ontology Categories:

          Function(s) Process(es)

          superoxide metabolic process
          response to oxidative stress
          aging
          response to nutrient
          response to organic cyclic compound
          removal of superoxide radicals
          response to estradiol
          negative regulation of catalytic activity
          positive regulation of neuron apoptotic process
          response to ethanol
          oxidation-reduction process
          NAD(P)H dehydrogenase (quinone) activity
          superoxide dismutase activity
          identical protein binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P005208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 126059 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89234 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P005208 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P005208 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P005208 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P476733 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98981 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10810871 - -
          Affymetrix Rat Gene 1.0 ST Array 10810868 - -
          Affymetrix Rat Gene 1.0 ST Array 10810870 - -
          Affymetrix Rat Gene 1.0 ST Array 10810874 - -
          Affymetrix Rat Gene 1.0 ST Array 10810869 - -
          Affymetrix Rat Gene 1.0 ST Array 10810873 - -
          Affymetrix Rat Gene 1.0 ST Array 10810872 - -
          Affymetrix Rat Gene 1.1 ST Array 10810867 - -
          Affymetrix Rat Clariom S Assay PSR1900004516.rn.1 TC1900000518.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900004515.rn.1 TC1900000518.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10810867 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21287126 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_2039396 - -
          RatRef-12 Expression BeadChip ILMN_58987 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline