Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00567167_m1
          See other HSD11B1 GE Assays ›
          Gene Symbol:
          Hsd11b1
          Gene Name:
          hydroxysteroid 11-beta dehydrogenase 1
          Gene Aliases:
          LRRGT00065
          Chromosome Location:
          Chr.13: 111946626 - 111996531 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00567167_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hsd11b1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017080.2 NP_058776.2 4 - 5 842 80
          XM_006250471.2 XP_006250533.1 5 - 6 623 80
          XM_006250473.2 XP_006250535.1 5 - 6 658 80
          GenBank mRNA AF542063.1 - 3 - 4 442 80
          BC078865.1 - 4 - 5 663 7109256 80
          FQ209646.1 - 4 - 5 609 80
          FQ209829.1 - 4 - 5 609 80
          FQ210302.1 - 4 - 5 607 80
          FQ210563.1 - 4 - 5 607 80
          FQ210596.1 - 4 - 5 609 80
          FQ211059.1 - 4 - 5 606 80
          FQ211404.1 - 4 - 5 608 80
          FQ212763.1 - 4 - 5 610 80
          FQ214152.1 - 4 - 5 609 80
          FQ214397.1 - 5 - 6 665 80
          FQ218558.1 - 4 - 5 607 80
          FQ218584.1 - 4 - 5 609 80
          FQ219123.1 - 4 - 5 609 80
          FQ219241.1 - 4 - 5 608 80
          FQ219385.1 - 4 - 5 607 80
          FQ219394.1 - 4 - 5 607 80
          FQ219517.1 - 4 - 5 609 80
          FQ219567.1 - 4 - 5 618 80
          FQ219624.1 - 4 - 5 542 80
          FQ220122.1 - 4 - 5 607 80
          J05107.1 - 4 - 5 590 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300002245.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus Rat Androgen Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hsd11b1

          Panther Classification:

          Molecular Function -

          dehydrogenase

          Gene Ontology Categories:

          Function(s) Process(es)

          glucocorticoid biosynthetic process
          glucocorticoid catabolic process
          metabolic process
          phthalate metabolic process
          lung development
          response to nutrient levels
          regulation of pentose-phosphate shunt
          oxidation-reduction process
          cellular response to estradiol stimulus
          11-beta-hydroxysteroid dehydrogenase [NAD(P)] activity
          steroid binding
          oxidoreductase activity
          NADP binding
          11-beta-hydroxysteroid dehydrogenase (NADP+) activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1018957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603850 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 2746 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 105518 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3877 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 51796 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1018957 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1018957 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1018957 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P426345 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1018957 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19672 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65967 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29308 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63228 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42722 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10770803 - -
          Affymetrix Rat Gene 1.0 ST Array 10770801 - -
          Affymetrix Rat Gene 1.0 ST Array 10770806 - -
          Affymetrix Rat Gene 1.0 ST Array 10770798 - -
          Affymetrix Rat Gene 1.0 ST Array 10770804 - -
          Affymetrix Rat Gene 1.0 ST Array 10770802 - -
          Affymetrix Rat Gene 1.0 ST Array 10770805 - -
          Affymetrix Rat Gene 1.0 ST Array 10770800 - -
          Affymetrix Rat Gene 1.0 ST Array 10770797 - -
          Affymetrix Rat Gene 1.1 ST Array 10770795 - -
          Rat Genome 230 2.0 Array 1386953_at - -
          Rat Genome 230 2.0 Array 1386953_PM_at - -
          Affymetrix HT Rat Focus Array 1386953_PM_at - -
          Affymetrix HT Rat Focus Array 1386953_at - -
          Affymetrix Rat Clariom S Assay PSR1300016028.rn.1 TC1300002245.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300016026.rn.1 TC1300002245.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300016030.rn.1 TC1300002245.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300016032.rn.1 TC1300002245.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300016033.rn.1 TC1300002245.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300016025.rn.1 TC1300002245.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300016036.rn.1 TC1300002245.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300016034.rn.1 TC1300002245.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300016038.rn.1 TC1300002245.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300016037.rn.1 TC1300002245.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300016031.rn.1 TC1300002245.rn.1 RTA TC
          Rat Expression Set 230 1386953_PM_at - -
          Rat Expression Set 230 1386953_at - -
          Affymetrix Rat Gene 1.0 ST Array 10770795 - -
          Rat Genome U34 Set rc_AI105448_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21812929 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369135 - -
          RatRef-12 Expression BeadChip ILMN_52838 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline