Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00568087_m1
          See other SLC3A1 GE Assays ›
          Gene Symbol:
          Slc3a1
          Gene Name:
          solute carrier family 3, member 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 8284937 - 8318649 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00568087_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc3a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017216.1 NP_058912.1 7 - 8 1363 59
          GenBank mRNA BC078852.1 - 7 - 8 1358 7110514 59
          M77345.1 - 7 - 8 1363 59
          M80804.1 - 7 - 8 1389 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600000109.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc3a1

          Panther Classification:

          Molecular Function -

          amylase

          Gene Ontology Categories:

          Function(s) Process(es)

          carbohydrate metabolic process
          amino acid transport
          catalytic activity
          cation binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P074875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880551 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106545 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 651834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 815199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216373 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33412 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 148662 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157255 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 169000 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P074875 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P074875 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P074875 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P421514 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73467 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73730 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5300 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10882676 - -
          Affymetrix Rat Gene 1.0 ST Array 10882680 - -
          Affymetrix Rat Gene 1.0 ST Array 10882678 - -
          Affymetrix Rat Gene 1.0 ST Array 10882684 - -
          Affymetrix Rat Gene 1.0 ST Array 10882677 - -
          Affymetrix Rat Gene 1.0 ST Array 10882682 - -
          Affymetrix Rat Gene 1.0 ST Array 10882681 - -
          Affymetrix Rat Gene 1.0 ST Array 10882679 - -
          Affymetrix Rat Gene 1.0 ST Array 10882675 - -
          Affymetrix Rat Gene 1.0 ST Array 10882674 - -
          Affymetrix Rat Gene 1.1 ST Array 10882672 - -
          Rat Genome 230 2.0 Array 1375182_PM_at - -
          Rat Genome 230 2.0 Array 1368440_PM_at - -
          Rat Genome 230 2.0 Array 1375182_at - -
          Rat Genome 230 2.0 Array 1368440_at - -
          Affymetrix HT Rat Focus Array 1368440_PM_at - -
          Affymetrix HT Rat Focus Array 1368440_at - -
          Affymetrix Rat Clariom S Assay PSR0600000681.rn.1 TC0600000109.rn.1 RTA TC
          Rat Expression Set 230 1368440_at - -
          Rat Expression Set 230 1368440_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10882672 - -
          Rat Genome U34 Set M80804_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21458812 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355705 - -
          RatRef-12 Expression BeadChip ILMN_51786 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline