Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00569290_m1
          See other PLAUR GE Assays ›
          Gene Symbol:
          Plaur
          Gene Name:
          plasminogen activator, urokinase receptor
          Gene Aliases:
          Par, Plaur3, uPAR, uPAR-2, uPAR-3
          Chromosome Location:
          Chr.1: 81328171 - 81344954 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00569290_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Plaur
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017350.1 NP_059046.1 2 - 3 189 86
          NM_134352.1 NP_599179.1 2 - 3 194 86
          GenBank mRNA AF007789.1 - 2 - 3 181 86
          BC127499.1 - 2 - 3 189 7380245 86
          FQ220340.1 - 2 - 3 202 86
          X71898.1 - 2 - 3 194 86
          X71899.1 - 2 - 3 189 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100001041.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Plaur

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          spermatogenesis
          positive regulation of gene expression
          urokinase plasminogen activator signaling pathway
          negative regulation of apoptotic process
          skeletal muscle tissue regeneration
          mesenchymal cell differentiation
          epithelial cell differentiation involved in prostate gland development
          receptor binding
          enzyme binding
          protein domain specific binding
          urokinase plasminogen activator receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P468141 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P038008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 670716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676308 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71175 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P468141 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P038008 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P038008 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P468141 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P038008 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P468141 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P468141 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702211768 - -
          Agilent Rat cDNA Microarray (G4105A) 702211768 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10704958 - -
          Affymetrix Rat Gene 1.0 ST Array 10704959 - -
          Affymetrix Rat Gene 1.0 ST Array 10704957 - -
          Affymetrix Rat Gene 1.1 ST Array 10704956 - -
          Rat Genome 230 2.0 Array 1395617_PM_at - -
          Rat Genome 230 2.0 Array 1395617_at - -
          Affymetrix Rat Clariom S Assay PSR0100007337.rn.1 TC0100001041.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100007336.rn.1 TC0100001041.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100007341.rn.1 TC0100001041.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100007338.rn.1 TC0100001041.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100007339.rn.1 TC0100001041.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100007340.rn.1 TC0100001041.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10704956 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21139516 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline