Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00569786_m1
          See other SPTBN2 GE Assays ›
          Gene Symbol:
          Sptbn2
          Gene Name:
          spectrin, beta, non-erythrocytic 2
          Gene Aliases:
          Spnb3
          Chromosome Location:
          Chr.1: 219964429 - 220006802 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00569786_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sptbn2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019167.1 NP_062040.1 19 - 20 4407 88
          XM_008760092.1 XP_008758314.1 18 - 19 4639 88
          XM_008760093.1 XP_008758315.1 18 - 19 4639 88
          XM_008760094.1 XP_008758316.1 19 - 20 4136 88
          GenBank mRNA AB001347.1 - 19 - 20 4407 88
          AB008551.1 - 18 - 19 4016 88
          AF225960.1 - 18 - 19 4016 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100003009.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sptbn2

          Gene Ontology Categories:

          Function(s) Process(es)

          synapse assembly
          synaptic vesicle exocytosis
          synaptic vesicle docking
          vesicle-mediated transport
          regulation of calcium ion-dependent exocytosis
          cerebellar Purkinje cell layer morphogenesis
          adult behavior
          multicellular organism growth
          actin filament capping
          actin binding
          structural constituent of cytoskeleton
          phospholipid binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P125013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 322859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482888 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897057 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345018 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82630 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109564 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172205 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49180 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P125013 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P125013 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P125013 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P596024 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700940773 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28939 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23345 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28944 - -
          Agilent Rat cDNA Microarray (G4105A) 700940773 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10712900 - -
          Affymetrix Rat Gene 1.0 ST Array 10712926 - -
          Affymetrix Rat Gene 1.0 ST Array 10712910 - -
          Affymetrix Rat Gene 1.0 ST Array 10712909 - -
          Affymetrix Rat Gene 1.0 ST Array 10712902 - -
          Affymetrix Rat Gene 1.0 ST Array 10712904 - -
          Affymetrix Rat Gene 1.0 ST Array 10712915 - -
          Affymetrix Rat Gene 1.0 ST Array 10712925 - -
          Affymetrix Rat Gene 1.0 ST Array 10712898 - -
          Affymetrix Rat Gene 1.0 ST Array 10712894 - -
          Affymetrix Rat Gene 1.0 ST Array 10712923 - -
          Affymetrix Rat Gene 1.0 ST Array 10712895 - -
          Affymetrix Rat Gene 1.0 ST Array 10712924 - -
          Affymetrix Rat Gene 1.0 ST Array 10712911 - -
          Affymetrix Rat Gene 1.0 ST Array 10712890 - -
          Affymetrix Rat Gene 1.0 ST Array 10712892 - -
          Affymetrix Rat Gene 1.0 ST Array 10712901 - -
          Affymetrix Rat Gene 1.0 ST Array 10712896 - -
          Affymetrix Rat Gene 1.0 ST Array 10712917 - -
          Affymetrix Rat Gene 1.0 ST Array 10712927 - -
          Affymetrix Rat Gene 1.0 ST Array 10712920 - -
          Affymetrix Rat Gene 1.0 ST Array 10712918 - -
          Affymetrix Rat Gene 1.0 ST Array 10712907 - -
          Affymetrix Rat Gene 1.0 ST Array 10712916 - -
          Affymetrix Rat Gene 1.0 ST Array 10712914 - -
          Affymetrix Rat Gene 1.0 ST Array 10712906 - -
          Affymetrix Rat Gene 1.0 ST Array 10712913 - -
          Affymetrix Rat Gene 1.0 ST Array 10712891 - -
          Affymetrix Rat Gene 1.0 ST Array 10712908 - -
          Affymetrix Rat Gene 1.0 ST Array 10712921 - -
          Affymetrix Rat Gene 1.0 ST Array 10712897 - -
          Affymetrix Rat Gene 1.0 ST Array 10712919 - -
          Affymetrix Rat Gene 1.0 ST Array 10712893 - -
          Affymetrix Rat Gene 1.0 ST Array 10712903 - -
          Affymetrix Rat Gene 1.0 ST Array 10712905 - -
          Affymetrix Rat Gene 1.0 ST Array 10712922 - -
          Affymetrix Rat Gene 1.0 ST Array 10712912 - -
          Affymetrix Rat Gene 1.0 ST Array 10712899 - -
          Affymetrix Rat Gene 1.1 ST Array 10712889 - -
          Rat Genome 230 2.0 Array 1367812_at - -
          Rat Genome 230 2.0 Array 1367812_PM_at - -
          Affymetrix HT Rat Focus Array 1367812_PM_at - -
          Affymetrix HT Rat Focus Array 1367812_at - -
          Affymetrix Rat Clariom S Assay PSR0100024378.rn.1 TC0100003009.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100024377.rn.1 TC0100003009.rn.1 RTA TC
          Rat Expression Set 230 1367812_PM_at - -
          Rat Expression Set 230 1367812_at - -
          Affymetrix Rat Gene 1.0 ST Array 10712889 - -
          Rat Genome U34 Set AB001347_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21313981 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366935 - -
          RatRef-12 Expression BeadChip ILMN_67122 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline