Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00569911_m1
          See other TEF GE Assays ›
          Gene Symbol:
          Tef
          Gene Name:
          thyrotrophic embryonic factor
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 123033961 - 123058606 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00569911_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tef
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019194.2 NP_062067.2 2 - 3 556 79
          XM_006242068.2 XP_006242130.1 2 - 3 625 79
          GenBank mRNA BC101895.1 - 2 - 3 556 7932583 79
          S58745.1 - 2 - 3 379 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700001507.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tef

          Panther Classification:

          Molecular Function -

          basic leucine zipper transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          rhythmic process
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          double-stranded DNA binding
          protein homodimerization activity
          sequence-specific DNA binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P788480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94074 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138909 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106530 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P788480 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P788480 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P788480 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15280 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18307 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104180 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10897999 - -
          Affymetrix Rat Gene 1.0 ST Array 10897998 - -
          Affymetrix Rat Gene 1.0 ST Array 10898002 - -
          Affymetrix Rat Gene 1.0 ST Array 10898000 - -
          Affymetrix Rat Gene 1.0 ST Array 10898001 - -
          Affymetrix Rat Gene 1.1 ST Array 10897997 - -
          Rat Genome 230 2.0 Array 1390819_at - -
          Rat Genome 230 2.0 Array 1390819_PM_at - -
          Rat Genome 230 2.0 Array 1369919_at - -
          Rat Genome 230 2.0 Array 1369919_PM_at - -
          Affymetrix HT Rat Focus Array 1369919_PM_at - -
          Affymetrix HT Rat Focus Array 1369919_at - -
          Affymetrix HT Rat Focus Array 1390819_PM_at - -
          Affymetrix HT Rat Focus Array 1390819_at - -
          Affymetrix Rat Clariom S Assay PSR0700011470.rn.1 TC0700001507.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700011471.rn.1 TC0700001507.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700011469.rn.1 TC0700001507.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700011473.rn.1 TC0700001507.rn.1 RTA TC
          Rat Expression Set 230 1369919_at - -
          Rat Expression Set 230 1369919_PM_at - -
          Rat Expression Set 230 1390819_PM_at - -
          Rat Expression Set 230 1390819_at - -
          Affymetrix Rat Gene 1.0 ST Array 10897997 - -
          Rat Genome U34 Set S58745_at - -
          Rat Genome U34 Set rc_AA998752_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21435654 - -
          ABI Rat Genome Survey Microarray 22009338 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372235 - -
          RatRef-12 Expression BeadChip ILMN_62721 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline