Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00570713_m1
          See other CD5 GE Assays ›
          Gene Symbol:
          Cd5
          Gene Name:
          Cd5 molecule
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 226770912 - 226791861 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00570713_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cd5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019295.1 NP_062168.1 1 - 2 145 82
          GenBank mRNA BC098918.1 - 1 - 2 116 7384975 118
          D10728.1 - 1 - 2 145 82
          X78985.1 - 1 - 2 56 118

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100007263.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cd5

          Gene Ontology Categories:

          Function(s) Process(es)

          receptor-mediated endocytosis
          regulation of signal transduction
          T cell costimulation
          apoptotic signaling pathway
          glycoprotein binding
          scavenger receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P409803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911651 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619888 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132755 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731463 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176520 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67127 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P409803 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P409803 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P409803 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P409803 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700531314 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23090 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67631 - -
          Agilent Rat cDNA Microarray (G4105A) 700531314 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10728767 - -
          Affymetrix Rat Gene 1.0 ST Array 10728777 - -
          Affymetrix Rat Gene 1.0 ST Array 10728769 - -
          Affymetrix Rat Gene 1.0 ST Array 10728768 - -
          Affymetrix Rat Gene 1.0 ST Array 10728773 - -
          Affymetrix Rat Gene 1.0 ST Array 10728774 - -
          Affymetrix Rat Gene 1.0 ST Array 10728771 - -
          Affymetrix Rat Gene 1.0 ST Array 10728778 - -
          Affymetrix Rat Gene 1.0 ST Array 10728776 - -
          Affymetrix Rat Gene 1.0 ST Array 10728779 - -
          Affymetrix Rat Gene 1.0 ST Array 10728775 - -
          Affymetrix Rat Gene 1.0 ST Array 10728770 - -
          Affymetrix Rat Gene 1.0 ST Array 10728772 - -
          Affymetrix Rat Gene 1.1 ST Array 10728766 - -
          Rat Genome 230 2.0 Array 1369025_at - -
          Rat Genome 230 2.0 Array 1369025_PM_at - -
          Affymetrix HT Rat Focus Array 1369025_PM_at - -
          Affymetrix HT Rat Focus Array 1369025_at - -
          Affymetrix Rat Clariom S Assay PSR0100058773.rn.1 TC0100007263.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100058777.rn.1 TC0100007263.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100058776.rn.1 TC0100007263.rn.1 RTA TC
          Rat Expression Set 230 1369025_at - -
          Rat Expression Set 230 1369025_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10728766 - -
          Rat Genome U34 Set D10728_at - -
          Rat Genome U34 Set X78985cds_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21103539 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357721 - -
          RatRef-12 Expression BeadChip ILMN_54670 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline