Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00570893_m1
          See other ITPKB GE Assays ›
          Gene Symbol:
          Itpkb
          Gene Name:
          inositol-trisphosphate 3-kinase B
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 98615287 - 98708150 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00570893_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Itpkb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019312.2 NP_062185.2 2 - 3 2388 68
          GenBank mRNA AJ242781.1 - 1937 68
          X74227.1 - 1711 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300000901.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Itpkb

          Panther Classification:

          Molecular Function -

          kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          cell surface receptor signaling pathway
          phosphorylation
          T cell differentiation
          positive thymic T cell selection
          thymic T cell selection
          positive regulation of Ras protein signal transduction
          positive regulation of alpha-beta T cell differentiation
          calmodulin binding
          ATP binding
          inositol-1,4,5-trisphosphate 3-kinase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P104548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272109 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601656 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310762 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32098 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53400 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103481 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165642 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134579 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50094 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 28143 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22366 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53214 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103123 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99496 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31724 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P104548 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P104548 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P104548 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P284561 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P255125 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6004 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87956 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49008 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74831 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41442 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58896 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7430 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98598 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10766166 - -
          Affymetrix Rat Gene 1.0 ST Array 10766169 - -
          Affymetrix Rat Gene 1.0 ST Array 10766162 - -
          Affymetrix Rat Gene 1.0 ST Array 10766167 - -
          Affymetrix Rat Gene 1.0 ST Array 10766164 - -
          Affymetrix Rat Gene 1.0 ST Array 10766168 - -
          Affymetrix Rat Gene 1.0 ST Array 10766165 - -
          Affymetrix Rat Gene 1.0 ST Array 10766163 - -
          Affymetrix Rat Gene 1.1 ST Array 10766161 - -
          Rat Genome 230 2.0 Array 1377984_PM_at - -
          Rat Genome 230 2.0 Array 1377984_at - -
          Rat Genome 230 2.0 Array 1369757_at - -
          Rat Genome 230 2.0 Array 1369757_PM_at - -
          Affymetrix HT Rat Focus Array 1377984_PM_at - -
          Affymetrix HT Rat Focus Array 1377984_at - -
          Affymetrix HT Rat Focus Array 1369757_at - -
          Affymetrix HT Rat Focus Array 1369757_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1300006334.rn.1 TC1300000901.rn.1 RTA TC
          Rat Expression Set 230 1369757_at - -
          Rat Expression Set 230 1369757_PM_at - -
          Rat Expression Set 230 1377984_PM_at - -
          Rat Expression Set 230 1377984_at - -
          Affymetrix Rat Gene 1.0 ST Array 10766161 - -
          Rat Genome U34 Set X74227cds_at - -
          Rat Genome U34 Set rc_AI102896_at - -
          Rat Genome U34 Set X74227cds_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21918605 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359923 - -
          RatRef-12 Expression BeadChip ILMN_65284 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline