Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00571516_m1
          See other ANXA2 GE Assays ›
          Gene Symbol:
          Anxa2
          Gene Name:
          annexin A2
          Gene Aliases:
          ANX2
          Chromosome Location:
          Chr.8: 75687134 - 75723589 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00571516_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Anxa2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019905.1 NP_063970.1 7 - 8 571 78
          GenBank mRNA BC059136.1 - 7 - 8 590 6920034 78
          FQ219918.1 - 2 - 3 137 78
          FQ225353.1 - 4 - 5 359 78
          L13039.1 - 6 - 7 542 78
          S73559.1 - 7 - 8 573 78
          X66871.1 - 7 - 8 571 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800001028.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Anxa2

          Panther Classification:

          Molecular Function -

          calcium-binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          membrane raft assembly
          positive regulation of protein phosphorylation
          membrane budding
          body fluid secretion
          collagen fibril organization
          positive regulation of vesicle fusion
          osteoclast development
          fibrinolysis
          negative regulation of catalytic activity
          positive regulation of fibroblast proliferation
          positive regulation of binding
          protein heterotetramerization
          regulation of fibrinolysis
          protein targeting to plasma membrane
          calcium ion binding
          calcium-dependent phospholipid binding
          phosphatidylinositol-4,5-bisphosphate binding
          Rab GTPase binding
          phospholipase A2 inhibitor activity
          S100 protein binding
          poly(A) RNA binding
          calcium-dependent protein binding
          actin filament binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P005576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604360 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 251910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 451533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308045 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162659 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75238 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166109 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90612 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150604 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P005576 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P005576 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P005576 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P532103 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700370911 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10732 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48847 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79966 - -
          Agilent Rat cDNA Microarray (G4105A) 700370911 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10911293 - -
          Affymetrix Rat Gene 1.0 ST Array 10911288 - -
          Affymetrix Rat Gene 1.0 ST Array 10911291 - -
          Affymetrix Rat Gene 1.0 ST Array 10911294 - -
          Affymetrix Rat Gene 1.0 ST Array 10911289 - -
          Affymetrix Rat Gene 1.0 ST Array 10911295 - -
          Affymetrix Rat Gene 1.0 ST Array 10911292 - -
          Affymetrix Rat Gene 1.0 ST Array 10911290 - -
          Affymetrix Rat Gene 1.1 ST Array 10911287 - -
          Rat Genome 230 2.0 Array 1367584_at - -
          Rat Genome 230 2.0 Array 1392250_at - -
          Rat Genome 230 2.0 Array 1392250_PM_at - -
          Rat Genome 230 2.0 Array 1367584_PM_at - -
          Affymetrix HT Rat Focus Array 1367584_PM_at - -
          Affymetrix HT Rat Focus Array 1367584_at - -
          Affymetrix HT Rat Focus Array 1392250_PM_at - -
          Affymetrix HT Rat Focus Array 1392250_at - -
          Affymetrix Rat Clariom S Assay PSR0800007431.rn.1 TC0800001028.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800007432.rn.1 TC0800001028.rn.1 RTA TC
          Rat Expression Set 230 1367584_at - -
          Rat Expression Set 230 1367584_PM_at - -
          Rat Expression Set 230 1392250_at - -
          Rat Expression Set 230 1392250_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10911287 - -
          Rat Genome U34 Set L13039_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22147762 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364260 - -
          RatRef-12 Expression BeadChip ILMN_57815 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline