Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00571769_m1
          See other SLC24A1 GE Assays ›
          Gene Symbol:
          Slc24a1
          Gene Name:
          solute carrier family 24 (sodium/potassium/calcium exchanger), member 1
          Gene Aliases:
          Nckx1
          Chromosome Location:
          Chr.8: 70411186 - 70436337 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00571769_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc24a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020090.1 NP_064475.1 5 - 6 2134 65
          GenBank mRNA AF176688.1 - 5 - 6 2134 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800002771.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Transporter Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc24a1

          Panther Classification:

          Molecular Function -

          secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          calcium ion transport
          visual perception
          sodium ion transmembrane transport
          calcium ion transmembrane transport
          calcium, potassium:sodium antiporter activity
          symporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P092058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 743530 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3075 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128558 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55015 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P092058 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P092058 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P092058 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P437973 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49875 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37822 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7485 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10918255 - -
          Affymetrix Rat Gene 1.0 ST Array 10918256 - -
          Affymetrix Rat Gene 1.0 ST Array 10918251 - -
          Affymetrix Rat Gene 1.0 ST Array 10918250 - -
          Affymetrix Rat Gene 1.0 ST Array 10918257 - -
          Affymetrix Rat Gene 1.0 ST Array 10918249 - -
          Affymetrix Rat Gene 1.0 ST Array 10918253 - -
          Affymetrix Rat Gene 1.0 ST Array 10918254 - -
          Affymetrix Rat Gene 1.0 ST Array 10918252 - -
          Affymetrix Rat Gene 1.1 ST Array 10918248 - -
          Rat Genome 230 2.0 Array 1387829_PM_at - -
          Rat Genome 230 2.0 Array 1387829_at - -
          Affymetrix HT Rat Focus Array 1387829_PM_at - -
          Affymetrix HT Rat Focus Array 1387829_at - -
          Affymetrix Rat Clariom S Assay PSR0800022501.rn.1 TC0800002771.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800022500.rn.1 TC0800002771.rn.1 RTA TC
          Rat Expression Set 230 1387829_PM_at - -
          Rat Expression Set 230 1387829_at - -
          Affymetrix Rat Gene 1.0 ST Array 10918248 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21528201 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline