Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00571968_m1
          See other TMEM27 GE Assays ›
          Gene Symbol:
          Tmem27
          Gene Name:
          transmembrane protein 27
          Gene Aliases:
          NX-17, Nx17, collectrin
          Chromosome Location:
          Chr.X: 32118082 - 32153687 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00571968_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem27
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020976.1 NP_066125.1 2 - 3 125 77
          GenBank mRNA AF178086.1 - 2 - 3 125 77
          BC078838.1 - 2 - 3 182 7129929 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00001639.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmem27

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          positive regulation of SNARE complex assembly
          positive regulation of insulin secretion involved in cellular response to glucose stimulus
          positive regulation of calcium ion-dependent exocytosis
          positive regulation of amino acid transport
          metallopeptidase activity
          peptidyl-dipeptidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P130469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 694409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 102079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444697 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 18758 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92916 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3767 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132360 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P130469 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P130469 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P130469 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P515580 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701869983 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5923 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32952 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70234 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5337 - -
          Agilent Rat cDNA Microarray (G4105A) 701869983 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10937764 - -
          Affymetrix Rat Gene 1.0 ST Array 10937767 - -
          Affymetrix Rat Gene 1.0 ST Array 10937768 - -
          Affymetrix Rat Gene 1.0 ST Array 10937766 - -
          Affymetrix Rat Gene 1.0 ST Array 10937763 - -
          Affymetrix Rat Gene 1.0 ST Array 10937765 - -
          Affymetrix Rat Gene 1.1 ST Array 10937762 - -
          Rat Genome 230 2.0 Array 1387013_PM_at - -
          Rat Genome 230 2.0 Array 1387013_at - -
          Affymetrix HT Rat Focus Array 1387013_at - -
          Affymetrix HT Rat Focus Array 1387013_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0X00010637.rn.1 TC0X00001639.rn.1 RTA TC
          Rat Expression Set 230 1387013_PM_at - -
          Rat Expression Set 230 1387013_at - -
          Affymetrix Rat Gene 1.0 ST Array 10937762 - -
          Rat Genome U34 Set rc_AA893216_g_at - -
          Rat Genome U34 Set rc_AI105157_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21330960 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366641 - -
          RatRef-12 Expression BeadChip ILMN_55252 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline