Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00572183_m1
          See other DIO1 GE Assays ›
          Gene Symbol:
          Dio1
          Gene Name:
          deiodinase, iodothyronine, type I
          Gene Aliases:
          5DI, ITDI1, TXDI1
          Chromosome Location:
          Chr.5: 126895286 - 126911520 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00572183_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dio1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021653.3 NP_067685.5 2 - 3 484 83
          GenBank mRNA BC083557.1 - 2 - 3 479 7110283 83
          CK480517.1 - 2 - 3 484 83
          X57999.1 - 2 - 3 483 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500003305.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dio1

          Gene Ontology Categories:

          Function(s) Process(es)

          generation of precursor metabolites and energy
          hormone biosynthetic process
          oxidation-reduction process
          thyroxine 5'-deiodinase activity
          selenium binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P155193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187956 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7665 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178911 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170676 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P155193 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P155193 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P155193 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P470649 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P194274 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701194078 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39556 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68565 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98482 - -
          Agilent Rat cDNA Microarray (G4105A) 701194078 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10878553 - -
          Affymetrix Rat Gene 1.0 ST Array 10878551 - -
          Affymetrix Rat Gene 1.0 ST Array 10878549 - -
          Affymetrix Rat Gene 1.0 ST Array 10878552 - -
          Affymetrix Rat Gene 1.0 ST Array 10878550 - -
          Affymetrix Rat Gene 1.1 ST Array 10878548 - -
          Rat Genome 230 2.0 Array 1369259_PM_at - -
          Rat Genome 230 2.0 Array 1369259_at - -
          Affymetrix HT Rat Focus Array 1369259_PM_at - -
          Affymetrix HT Rat Focus Array 1369259_at - -
          Affymetrix Rat Clariom S Assay PSR0500024010.rn.1 TC0500003305.rn.1 RTA TC
          Rat Expression Set 230 1369259_at - -
          Rat Expression Set 230 1369259_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10878548 - -
          Rat Genome U34 Set rc_AI639418_at - -
          Rat Genome U34 Set X57999cds_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21920936 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350755 - -
          RatRef-12 Expression BeadChip ILMN_61587 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline