Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00572234_m1
          See other SYT7 GE Assays ›
          Gene Symbol:
          Syt7
          Gene Name:
          synaptotagmin VII
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 226435836 - 226498009 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00572234_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Syt7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021659.1 NP_067691.1 4 - 5 513 101
          XM_006231065.2 XP_006231127.1 8 - 9 1508 101
          XM_006231066.2 XP_006231128.1 7 - 8 1904 101
          XM_006231067.2 XP_006231129.1 7 - 8 1376 101
          XM_006231068.2 XP_006231130.1 7 - 8 1280 101
          XM_006231069.2 XP_006231131.1 5 - 6 788 101
          GenBank mRNA AF336852.1 - 9 - 10 1328 101
          AF336853.1 - 8 - 9 1100 101
          AF336854.1 - 4 - 5 353 101
          AF336855.1 - 8 - 9 1205 101
          AF336856.1 - 6 - 7 704 101
          AF336857.1 - 6 - 7 713 101
          AF336858.1 - 6 - 7 845 101
          AF336859.1 - 7 - 8 977 101
          AF336860.1 - 7 - 8 1073 101
          U20106.1 - 4 - 5 513 101

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100003196.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Syt7

          Panther Classification:

          Molecular Function -

          membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          plasma membrane repair
          detection of calcium ion
          exocytosis
          regulation of calcium ion-dependent exocytosis
          positive regulation of calcium ion-dependent exocytosis
          regulation of insulin secretion
          transporter activity
          calcium ion binding
          calcium-dependent phospholipid binding
          syntaxin binding
          clathrin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P042219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719179 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 743178 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 528020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382816 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384916 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615353 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52444 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 45250 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91817 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158108 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128667 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 148072 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17664 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P042219 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P042219 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P042219 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12088 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701914239 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49042 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25893 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60458 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42114 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75511 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75894 - -
          Agilent Rat cDNA Microarray (G4105A) 701914239 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10713886 - -
          Affymetrix Rat Gene 1.0 ST Array 10713878 - -
          Affymetrix Rat Gene 1.0 ST Array 10713891 - -
          Affymetrix Rat Gene 1.0 ST Array 10713882 - -
          Affymetrix Rat Gene 1.0 ST Array 10713887 - -
          Affymetrix Rat Gene 1.0 ST Array 10713893 - -
          Affymetrix Rat Gene 1.0 ST Array 10713879 - -
          Affymetrix Rat Gene 1.0 ST Array 10713880 - -
          Affymetrix Rat Gene 1.0 ST Array 10713881 - -
          Affymetrix Rat Gene 1.0 ST Array 10713889 - -
          Affymetrix Rat Gene 1.0 ST Array 10713883 - -
          Affymetrix Rat Gene 1.0 ST Array 10713877 - -
          Affymetrix Rat Gene 1.0 ST Array 10713888 - -
          Affymetrix Rat Gene 1.0 ST Array 10713885 - -
          Affymetrix Rat Gene 1.0 ST Array 10713884 - -
          Affymetrix Rat Gene 1.0 ST Array 10713892 - -
          Affymetrix Rat Gene 1.0 ST Array 10713890 - -
          Affymetrix Rat Gene 1.1 ST Array 10713876 - -
          Rat Genome 230 2.0 Array 1370515_x_at - -
          Rat Genome 230 2.0 Array 1370514_a_at - -
          Rat Genome 230 2.0 Array 1370515_PM_x_at - -
          Rat Genome 230 2.0 Array 1370514_PM_a_at - -
          Rat Genome 230 2.0 Array 1369185_a_at - -
          Rat Genome 230 2.0 Array 1394802_PM_at - -
          Rat Genome 230 2.0 Array 1369185_PM_a_at - -
          Rat Genome 230 2.0 Array 1394802_at - -
          Affymetrix HT Rat Focus Array 1370515_x_at - -
          Affymetrix HT Rat Focus Array 1370515_PM_x_at - -
          Affymetrix HT Rat Focus Array 1369185_PM_a_at - -
          Affymetrix HT Rat Focus Array 1370514_a_at - -
          Affymetrix HT Rat Focus Array 1370514_PM_a_at - -
          Affymetrix HT Rat Focus Array 1394802_at - -
          Affymetrix HT Rat Focus Array 1369185_a_at - -
          Affymetrix HT Rat Focus Array 1394802_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100026357.rn.1 TC0100003196.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100026375.rn.1 TC0100003196.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100026362.rn.1 TC0100003196.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100026373.rn.1 TC0100003196.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100026356.rn.1 TC0100003196.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100026355.rn.1 TC0100003196.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100026359.rn.1 TC0100003196.rn.1 RTA TC
          Rat Expression Set 230 1370514_a_at - -
          Rat Expression Set 230 1369185_a_at - -
          Rat Expression Set 230 1370515_PM_x_at - -
          Rat Expression Set 230 1370514_PM_a_at - -
          Rat Expression Set 230 1370515_x_at - -
          Rat Expression Set 230 1369185_PM_a_at - -
          Rat Expression Set 230 1394802_at - -
          Rat Expression Set 230 1394802_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10713876 - -
          Rat Genome U34 Set U20106_at - -
          Rat Genome U34 Set U20106_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21991231 - -
          ABI Rat Genome Survey Microarray 22364230 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_2039207 - -
          RatRef-12 Expression BeadChip ILMN_53108 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline