Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00573029_m1
          See other GABRA6 GE Assays ›
          Gene Symbol:
          Gabra6
          Gene Name:
          gamma-aminobutyric acid (GABA) A receptor, alpha 6
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 27847447 - 27862685 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00573029_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gabra6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021841.1 NP_068613.1 8 - 9 1083 144
          XM_006246128.2 XP_006246190.1 8 - 9 1232 144
          XM_008767636.1 XP_008765858.1 6 - 7 1154 144
          GenBank mRNA FQ213316.1 - 1 - 2 173 144

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000002602.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gabra6

          Panther Classification:

          Molecular Function -

          ligand-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          chloride transport
          gamma-aminobutyric acid signaling pathway
          ion transmembrane transport
          protein heterooligomerization
          synaptic transmission, GABAergic
          chloride transmembrane transport
          GABA-A receptor activity
          extracellular ligand-gated ion channel activity
          inhibitory extracellular ligand-gated ion channel activity
          chloride channel activity
          drug binding
          benzodiazepine receptor activity
          GABA-gated chloride ion channel activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P15548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128336 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 251245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316816 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414670 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701338 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 2468 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44874 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15548 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15548 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P15548 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15548 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63262 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75549 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10742187 - -
          Affymetrix Rat Gene 1.0 ST Array 10742188 - -
          Affymetrix Rat Gene 1.0 ST Array 10742191 - -
          Affymetrix Rat Gene 1.0 ST Array 10742184 - -
          Affymetrix Rat Gene 1.0 ST Array 10742190 - -
          Affymetrix Rat Gene 1.0 ST Array 10742186 - -
          Affymetrix Rat Gene 1.0 ST Array 10742185 - -
          Affymetrix Rat Gene 1.0 ST Array 10742189 - -
          Affymetrix Rat Gene 1.1 ST Array 10742183 - -
          Rat Genome 230 2.0 Array 1369861_at - -
          Rat Genome 230 2.0 Array 1369861_PM_at - -
          Affymetrix HT Rat Focus Array 1369861_at - -
          Affymetrix HT Rat Focus Array 1369861_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000023263.rn.1 TC1000002602.rn.1 RTA TC
          Rat Expression Set 230 1369861_PM_at - -
          Rat Expression Set 230 1369861_at - -
          Affymetrix Rat Gene 1.0 ST Array 10742183 - -
          Rat Genome U34 Set L08495cds_s_at - -
          Rat Genome U34 Set L08495cds_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21711022 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368214 - -
          RatRef-12 Expression BeadChip ILMN_63181 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline