Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00573576_m1
          See other INSRR GE Assays ›
          Gene Symbol:
          Insrr
          Gene Name:
          insulin receptor-related receptor
          Gene Aliases:
          Sirr
          Chromosome Location:
          Chr.2: 187161947 - 187181400 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00573576_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Insrr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022212.2 NP_071548.2 1 - 2 483 65
          XM_006232749.2 XP_006232811.2 1 - 2 553 65
          XM_008761207.1 XP_008759429.1 1 - 2 553 65
          GenBank mRNA D13965.1 - 1 - 2 85 65
          D13966.1 - 1 - 2 85 65
          M90661.1 - 1 - 2 483 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200001520.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Protein Kinase Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Insrr

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          transmembrane receptor protein tyrosine kinase signaling pathway
          peptidyl-tyrosine phosphorylation
          male sex determination
          actin cytoskeleton reorganization
          protein autophosphorylation
          cellular response to alkaline pH
          transmembrane receptor protein tyrosine kinase activity
          ATP binding
          phosphatidylinositol 3-kinase binding
          insulin receptor substrate binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P005346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781178 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772551 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 876019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311989 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170845 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58645 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37901 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P005346 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P005346 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P005346 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P491872 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P501086 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29519 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10816355 - -
          Affymetrix Rat Gene 1.0 ST Array 10816364 - -
          Affymetrix Rat Gene 1.0 ST Array 10816370 - -
          Affymetrix Rat Gene 1.0 ST Array 10816375 - -
          Affymetrix Rat Gene 1.0 ST Array 10816363 - -
          Affymetrix Rat Gene 1.0 ST Array 10816367 - -
          Affymetrix Rat Gene 1.0 ST Array 10816369 - -
          Affymetrix Rat Gene 1.0 ST Array 10816360 - -
          Affymetrix Rat Gene 1.0 ST Array 10816373 - -
          Affymetrix Rat Gene 1.0 ST Array 10816377 - -
          Affymetrix Rat Gene 1.0 ST Array 10816372 - -
          Affymetrix Rat Gene 1.0 ST Array 10816376 - -
          Affymetrix Rat Gene 1.0 ST Array 10816359 - -
          Affymetrix Rat Gene 1.0 ST Array 10816362 - -
          Affymetrix Rat Gene 1.0 ST Array 10816358 - -
          Affymetrix Rat Gene 1.0 ST Array 10816365 - -
          Affymetrix Rat Gene 1.0 ST Array 10816374 - -
          Affymetrix Rat Gene 1.0 ST Array 10816361 - -
          Affymetrix Rat Gene 1.0 ST Array 10816371 - -
          Affymetrix Rat Gene 1.0 ST Array 10816357 - -
          Affymetrix Rat Gene 1.0 ST Array 10816366 - -
          Affymetrix Rat Gene 1.0 ST Array 10816368 - -
          Affymetrix Rat Gene 1.0 ST Array 10816354 - -
          Affymetrix Rat Gene 1.0 ST Array 10816356 - -
          Affymetrix Rat Gene 1.1 ST Array 10824188 - -
          Affymetrix Rat Gene 1.1 ST Array 10816353 - -
          Rat Genome 230 2.0 Array 1369514_at - -
          Rat Genome 230 2.0 Array 1388065_at - -
          Rat Genome 230 2.0 Array 1369514_PM_at - -
          Rat Genome 230 2.0 Array 1369515_s_at - -
          Rat Genome 230 2.0 Array 1388065_PM_at - -
          Rat Genome 230 2.0 Array 1369515_PM_s_at - -
          Affymetrix HT Rat Focus Array 1369515_PM_s_at - -
          Affymetrix HT Rat Focus Array 1388065_at - -
          Affymetrix HT Rat Focus Array 1369514_PM_at - -
          Affymetrix HT Rat Focus Array 1388065_PM_at - -
          Affymetrix HT Rat Focus Array 1369515_s_at - -
          Affymetrix HT Rat Focus Array 1369514_at - -
          Affymetrix Rat Clariom S Assay PSR0200009417.rn.1 TC0200001520.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009418.rn.1 TC0200001520.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009416.rn.1 TC0200001520.rn.1 RTA TC
          Rat Expression Set 230 1369515_PM_s_at - -
          Rat Expression Set 230 1369515_s_at - -
          Rat Expression Set 230 1369514_at - -
          Rat Expression Set 230 1388065_PM_at - -
          Rat Expression Set 230 1369514_PM_at - -
          Rat Expression Set 230 1388065_at - -
          Affymetrix Rat Gene 1.0 ST Array 10816353 - -
          Rat Genome U34 Set M90660exon_g_at - -
          Rat Genome U34 Set D13965_s_at - -
          Rat Genome U34 Set D13966_at - -
          Rat Genome U34 Set M90660exon_at - -
          Rat Genome U34 Set M90661_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21210459 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350245 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline