Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00573954_m1
          See other PDCD4 GE Assays ›
          Gene Symbol:
          Pdcd4
          Gene Name:
          programmed cell death 4
          Gene Aliases:
          Dug
          Chromosome Location:
          Chr.1: 274625131 - 274648207 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00573954_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pdcd4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022265.2 NP_071601.2 4 - 5 552 69
          XM_006231613.2 XP_006231675.1 5 - 6 833 69
          XM_006231614.2 XP_006231676.1 5 - 6 885 69
          XM_006231615.2 XP_006231677.1 5 - 6 731 69
          XM_006231616.2 XP_006231678.1 5 - 6 708 69
          GenBank mRNA AF239739.2 - 4 - 5 552 69
          BC167751.1 - 5 - 6 740 9027822 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100003832.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pdcd4

          Panther Classification:

          Molecular Function -

          translation factor translational protein

          Gene Ontology Categories:

          Function(s) Process(es)

          apoptotic process
          cell aging
          positive regulation of smooth muscle cell apoptotic process
          negative regulation of apoptotic process
          negative regulation of JUN kinase activity
          negative regulation of transcription, DNA-templated
          regulation of protein metabolic process
          molecular_function
          RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919668 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964293 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115790 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 180812 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6682 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12191 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12191 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12191 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12191 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700493227 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35549 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86638 - -
          Agilent Rat cDNA Microarray (G4105A) 700493227 - -
          Agilent Rat cDNA Microarray (G4105A) 700233860 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10716144 - -
          Affymetrix Rat Gene 1.0 ST Array 10716139 - -
          Affymetrix Rat Gene 1.0 ST Array 10716145 - -
          Affymetrix Rat Gene 1.0 ST Array 10716137 - -
          Affymetrix Rat Gene 1.0 ST Array 10716138 - -
          Affymetrix Rat Gene 1.0 ST Array 10716146 - -
          Affymetrix Rat Gene 1.0 ST Array 10716142 - -
          Affymetrix Rat Gene 1.0 ST Array 10716147 - -
          Affymetrix Rat Gene 1.0 ST Array 10716143 - -
          Affymetrix Rat Gene 1.0 ST Array 10716140 - -
          Affymetrix Rat Gene 1.0 ST Array 10716141 - -
          Affymetrix Rat Gene 1.1 ST Array 10716136 - -
          Rat Genome 230 2.0 Array 1383327_at - -
          Rat Genome 230 2.0 Array 1383326_a_at - -
          Rat Genome 230 2.0 Array 1383328_PM_x_at - -
          Rat Genome 230 2.0 Array 1383327_PM_at - -
          Rat Genome 230 2.0 Array 1383328_x_at - -
          Rat Genome 230 2.0 Array 1387521_at - -
          Rat Genome 230 2.0 Array 1383329_at - -
          Rat Genome 230 2.0 Array 1383329_PM_at - -
          Rat Genome 230 2.0 Array 1383326_PM_a_at - -
          Rat Genome 230 2.0 Array 1387521_PM_at - -
          Affymetrix HT Rat Focus Array 1383326_a_at - -
          Affymetrix HT Rat Focus Array 1383328_x_at - -
          Affymetrix HT Rat Focus Array 1383329_at - -
          Affymetrix HT Rat Focus Array 1383327_at - -
          Affymetrix HT Rat Focus Array 1387521_PM_at - -
          Affymetrix HT Rat Focus Array 1383326_PM_a_at - -
          Affymetrix HT Rat Focus Array 1383328_PM_x_at - -
          Affymetrix HT Rat Focus Array 1383329_PM_at - -
          Affymetrix HT Rat Focus Array 1387521_at - -
          Affymetrix HT Rat Focus Array 1383327_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100031289.rn.1 TC0100003832.rn.1 RTA TC
          Rat Expression Set 230 1387521_PM_at - -
          Rat Expression Set 230 1387521_at - -
          Rat Expression Set 230 1383327_at - -
          Rat Expression Set 230 1383327_PM_at - -
          Rat Expression Set 230 1383326_a_at - -
          Rat Expression Set 230 1383329_at - -
          Rat Expression Set 230 1383328_PM_x_at - -
          Rat Expression Set 230 1383328_x_at - -
          Rat Expression Set 230 1383329_PM_at - -
          Rat Expression Set 230 1383326_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10716136 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21720956 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354540 - -
          RatRef-12 Expression BeadChip ILMN_68600 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline